Skip to Content
Merck
All Photos(1)

Key Documents

372439

Sigma-Aldrich

Perfluoro(methyldecalin), mixture of isomers

technical grade, 80%

Synonym(s):

Heptadecafluorodecahydro(trifluoromethyl)naphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11F20
CAS Number:
Molecular Weight:
512.09
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

80%

form

liquid

refractive index

n20/D 1.317 (lit.)

bp

137-160 °C (lit.)

mp

−40 °C (lit.)

density

1.95 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)C1(F)C(F)(F)C(F)(F)C2(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C2(F)C1(F)F.FC(F)(F)C3(F)C(F)(F)C(F)(F)C(F)(F)C4(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C34F

InChI

1S/C11F20/c12-1-2(13,6(19,20)10(27,28)9(25,26)4(1,15)16)5(17,18)8(23,24)7(21,22)3(1,14)11(29,30)31

InChI key

LWRNQOBXRHWPGE-UHFFFAOYSA-N

General description

Perfluoro(methyldecalin) is a fluorinated solvent.

Application

Perfluoro(methyldecalin is suitable reagent used to investigate the reactions of iodide anions and anions derived from glow discharge ionization of perfluoro(methyldecalin) with positive ions derived from poly(ethylene glycol). It is suitable reagent used for the oxidation of alcohols with the ruthenium porphyrin at higher temperatures (140ºC) and elevated oxygen pressures (50psi). Perfluoro(methyldecalin (Perfluoro-1-Methyldecalin) may be used in the preparation of undecafluoro-6-trifluoromethyltetralin via defluorination.

Other Notes

Contains perfluorodecalin

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.0 °F - closed cup

Flash Point(C)

112.80 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M K Bakulin et al.
Prikladnaia biokhimiia i mikrobiologiia, 43(4), 443-449 (2007-10-13)
The addition of perfluorocarbons (perfluorodecalin, carbogal, and perfluoromethyldecalin) to nitrogen-free liquid media during the submerged cultivation of bacteria of the genus Azotobacter was followed by (1) increases in biomass accumulation and nitrogenase activity and (2) fixation of molecular nitrogen. Addition
37. Polycyclic fluoroaromatic compounds. Part II. Perfluoromethylnaphthalenes and derived compounds.
Gething B, et al.
Journal of the Chemical Society, 190-193 (1962)
D J McIver et al.
Biochimica et biophysica acta, 751(1), 74-80 (1983-03-22)
An effective substitute for pulmonary surfactant must be able to reduce water-vapour surface tensions to under 1 mN/m and it must spread rapidly and spontaneously at the air interface from the aqueous phase. Pure dipalmitoylphosphatidylcholine meets the former requirement, but
Vasily N Korotchenko et al.
Organic & biomolecular chemistry, 6(11), 1961-1965 (2008-05-16)
Carbonylruthenium tetrakis(pentafluorophenyl)porphyrin Ru(TPFPP)(CO) was utilized for the aerobic oxidation of alcohols. The in situ activation of the catalyst with mCPBA provided a species capable of catalyzing the oxidation of alcohols with molecular oxygen. The choice of solvent and additive was
M K Bakulin et al.
Mikrobiologiia, 75(3), 371-376 (2006-07-29)
The isolates of pseudomonads (56) and of Plesiomonas shigelloides (7) exhibiting ice-forming activity were isolated from plant leaves and rhizosphere. The theoretical possibility of the application of perfluorocarbons (PFC) with a gas-transporting function (perfluorodecalin, carbogal, and perfluoromethyldecalin) for the cultivation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service