359238
Perfluorooctane
98%
Synonym(s):
Octadecafluorooctane
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
CF3(CF2)6CF3
CAS Number:
Molecular Weight:
438.06
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
form
liquid
refractive index
n20/D 1.3 (lit.)
bp
103-104 °C (lit.)
mp
−25 °C (lit.)
density
1.766 g/mL at 25 °C (lit.)
application(s)
PFAS testing
functional group
fluoro
SMILES string
FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
InChI
1S/C8F18/c9-1(10,3(13,14)5(17,18)7(21,22)23)2(11,12)4(15,16)6(19,20)8(24,25)26
InChI key
YVBBRRALBYAZBM-UHFFFAOYSA-N
General description
Perfluorooctane is a polyfluoroalkyl substance. Perfluorohexanoic acid (PFHxA), a 6-carbon perfluoroalkyl is an alternate of perfluorooctane. Phase behavior of semifluorinated n-alkanes in perfluorooctane has been studied .
Perfluorooctane, an oxygen carrier, is an alkane that is used as a solvent for dissolving gaseous solutes.
Perfluorooctane, an oxygen carrier, is an alkane that is used as a solvent for dissolving gaseous solutes.
Application
Perfluorooctane may be used in chemical synthesis.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Brett Drury et al.
The British journal of ophthalmology, 95(5), 694-698 (2010-10-12)
Inferior retinal detachment pathology can be difficult to manage due to inadequate tamponade with low specific gravity tamponade agents and the propensity for the development of proliferative vitreoretinopathy (PVR). We report the efficacy and adverse effects associated with the use
Cathrine Carlsen Bach et al.
Critical reviews in toxicology, 45(1), 53-67 (2014-11-06)
Exposure to perfluoroalkyl and polyfluoroalkyl substances (PFASs) is ubiquitous in most regions of the world. The most commonly studied PFASs are perfluorooctane sulfonate (PFOS) and perfluorooctanoate (PFOA). Animal studies indicate that maternal PFAS exposure is associated with reduced fetal growth.
Vicente Martínez-Castillo et al.
Retina (Philadelphia, Pa.), 32(7), 1350-1355 (2012-03-15)
To compare outcomes of eyes that did or did not develop an epiretinal membrane (ERM) after repair of primary pseudophakic or aphakic rhegmatogenous retinal detachment by pars plana vitrectomy alone. Interventional, prospective, consecutive case series. Of 312 consecutive eyes that
Dilraj S Grewal et al.
Ophthalmology, 126(10), 1399-1408 (2019-02-04)
To report the structural and functional outcomes of autologous neurosensory retinal transplant for closure of refractory large macular holes (MHs). Multicenter, retrospective, consecutive case series. A total of 41 eyes of 41 patients with a full-thickness MH refractory to prior
Aggregation of a semifluorinated n-alkane in perfluorooctane.
Lo Nostro P and Chen SH.
The Journal of Physical Chemistry, 97(24), 6535-6540 (1993)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service