Skip to Content
Merck
All Photos(3)

Key Documents

346462

Sigma-Aldrich

Zirconium(IV) oxynitrate hydrate

technical grade

Synonym(s):

Zirconyl nitrate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZrO(NO3)2 · xH2O
CAS Number:
Molecular Weight:
231.23 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
technical grade
form:
powder

grade

technical grade

form

powder

composition

Degree of hydration, ~6

concentration

25-30% Zr (after ignition to oxide, gravimetric)

application(s)

battery manufacturing

SMILES string

O=[Zr++].[H]O[H].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/2NO3.H2O.O.Zr/c2*2-1(3)4;;;/h;;1H2;;/q2*-1;;;+2

InChI key

BGMQNYCWALSARE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Zirconium(IV) oxynitrate hydrate is an inorganic salt primarily used as a precursor for the synthesis of zirconium nanoparticles, to fabricate Li-ion batteries, and as a catalyst in various chemical reactions.

Application

Zirconium(IV) oxynitrate hydrate can be used:
  • As a starting material to prepare electrolytes for Li-ion batteries.
  • For the surface modification of cathodes, to enhance the electrochemical properties of all-solid-state batteries.
  • As a precursor for surfactant-free synthesis of zirconia nanostructures.
It can also be used as a catalyst for:
  • Regiospecific alcoholysis and aminolysis of epoxides.
  • One-pot synthesis and oxidation of bis- and tris(indolyl)methanes into conjugated chromophores.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhanced electrochemical properties of all-solid-state batteries using a surface-modified LiNi 0.6 Co 0.2 Mn 0.2 O 2 Cathode
Chung Bum Lim, et al.
Journal of Electrochemical Science and Technology, 11, 411-420 (2020)
Mild regiospecific alcoholysis and aminolysis of epoxides catalyzed by zirconium(IV) oxynitrate
Sandip S. Shinde, et al.
Tetrahedron Letters, 56, 5916-5919 (2015)
Zirconia Nanostructures: Novel Facile Surfactant-Free Preparation and Characterization
Sahar Zinatloo-Ajabshir, et al.
International Journal of Applied Ceramic Technology, 13, 108-115 (2016)
The Structure and Electrical Properties of Li7La3Zr1.0Sn1.0O12 as Electrolyte Material for Li-ion Batteries
Siti Nur Adlina Norazman, et al.
Advances in Science and Technology, 118, 27-32 (2022)
Zirconium(IV)-catalyzed one-pot synthesis and oxidation of bis- and tris(indolyl)methanes into conjugated chromophores as new pH indicators or calorimetric chemosensors for transition metals
Alireza Khorshidi, et al.
Tetrahedron Letters, 55, 3873-3877 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service