Skip to Content
Merck
All Photos(3)

Documents

238554

Sigma-Aldrich

Lanthanum(III) nitrate hydrate

99.9% trace metals basis

Synonym(s):

Lanthanum nitrate hydrate, Lanthanum trinitrate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
La(NO3)3·xH2O
CAS Number:
Molecular Weight:
324.92 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

(powder, crystals or chunks)

composition

Degree of hydration, 3-5

reaction suitability

reagent type: catalyst
core: lanthanum

impurities

≤2000.0 ppm Trace Rare Earth Analysis

SMILES string

O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/La.3NO3.H2O/c;3*2-1(3)4;/h;;;;1H2/q+3;3*-1;

InChI key

HVMFKXBHFRRAAD-UHFFFAOYSA-N

General description

Lanthanum(III) nitrate hydrate is a white crystalline solid that is highly soluble in water. It is also utilized in various fields such as catalysts, ceramics, and as a source of lanthanum ions in chemical synthesis.

Application

Lanthanum(III) nitrate hydrate can be used:
  • As a starting material to synthesize lanthanum zirconate nanoparticles.
  • As a precursor to prepare luminescent lanthanum-tricarboxytriphenylamine (La-TCA) coordination polymer nanosheets.
  • To fabricate perovskite oxide-type LaFeO3 dendrites for chemical sensors.
  • To synthesize LaMnO3 perovskite catalysts by using citric acid sol–gel method and La0.6Sr0.4MnO3 perovskites via coprecipitation method. The LaMnO3 perovskite catalysts has used for the oxidation of propane for emission control of hydrocarbon volatile organic compounds.

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Precisely tailored LaFeO3 dendrites using urea and piperazine hexahydrate for the highly selective and sensitive detection of trace level acetone
K Palani Thiruppathi and Devaraj Nataraj
Materials Advances, 1, 2971-2982 (2020)
Jie Jian et al.
Nature communications, 10(1), 2609-2609 (2019-06-15)
Addressing the intrinsic charge transport limitation of metal oxides has been of significance for pursuing viable PEC water splitting photoelectrodes. Growing a photoelectrode with conductive nanoobjects embedded in the matrix is promising for enhanced charge transport but remains a challenge
Multi-responsive luminescent coordination polymer nanosheets for selective detection of nitroaromatics
Ting Pan, et al.
Chemical Communications (Cambridge, England), 58, 7809-7812 (2022)
Facile coprecipitation synthesis of La0.6Sr0.4MnO3 perovskites with high surface area
Fei Huang, et al.
Materials Letters, 210, 287-290 (2018)
Lanthanum zirconate nanoparticles and ceramics produced using a nitrate-modified alkoxide synthesis route
Elena-Daniela Ion, et al
Journal of Sol-Gel Science and Technology, 44, 203-209 (2007)

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service