Skip to Content
Merck
All Photos(3)

Key Documents

238538

Sigma-Aldrich

Cerium(III) nitrate hexahydrate

99% trace metals basis

Synonym(s):

Cerium trinitrate, Cerous nitrate hexahydrate, Nitric acid cerium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ce(NO3)3 · 6H2O
CAS Number:
Molecular Weight:
434.22
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99% trace metals basis
form:
crystals and lumps

Quality Level

Assay

99% trace metals basis

form

crystals and lumps

reaction suitability

reagent type: catalyst
core: cerium

impurities

1-2% La

SMILES string

[Ce+3].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Ce.3NO3.6H2O/c;3*2-1(3)4;;;;;;/h;;;;6*1H2/q+3;3*-1;;;;;;

InChI key

QQZMWMKOWKGPQY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cerium (III) nitrate hexahydrate is the most common form of cerium nitrate. It is a white-to-yellow crystalline salt, and it is highly soluble in water, alcohol, and acetone, although solutions can appear slightly hazy. Like many trivalent metal nitrates, cerium (III) nitrate hexahydrate has a low-melting point of only 57 °C and thermally decomposes at low temperatures too, beginning at 190 °C and proceeding rapidly at 280 °C with complete decomposition at 390-400 °C.

Application

Cerium (III) nitrate hexahydrate is widely used as a source of cerium, especially in the synthesis of micro- or nano-structured ceria (cerium oxide). Because of its high solubility and low decomposition temperature, cerium (III) nitrate hexahydrate is an ideal reagent for hydrothermal reactions, sol-gel processing, and co-precipitation and calcination reactions.

Ceria is one of the best studied materials-platforms for catalysis and solid oxide fuel cells. Consequently scientists have developed well-controlled synthetic protocols using cerium (III) nitrate hexahydrate for preparing ceria nanoparticles including nano cubes, nanotubes, and nanorods. The tailorable synthesis facilitates using ceria as a catalyst support, for example leveraging the porosity of the microstructure to impregnate metals and metal alloys, or for example synthesizing ceria-based materials like ceria-zirconia.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metal and ligand interaction of Lanthanides with coumarin derivatives. Part I. Complexation of 3-(1-Aminoethylidene)-2H-Chromene-2,4(3H)-Dione with La(III), Ce(III), Nd(III) and Ho(III)
Swiatek M,Kufelnicki A
Acta Poloniae Pharmaceutica, 69(6), 1000-1007 (2012)
Room temperature co-precipitation of nanocrystalline CeO2 and Ce0.8Gd0.2O1.9-d powder
Materials Letters, 61 , 1904-1907 (2007)
Ternary platinum?cobalt?indium nanoalloy on ceria as a highly efficient catalyst for the oxidative dehydrogenation of propane using CO2
Xing F, et al.
Nature catalysis, 55-65 (2022)
Kenji Kaneko et al.
Nano letters, 7(2), 421-425 (2007-02-15)
Colloidal cerium oxide (CeO2) nanocrystals prepared by hydrothermal synthesis were characterized by high-resolution transmission electron microscopy (HRTEM) and three-dimensional electron tomography (3D-ET). HRTEM images of individual CeO2 nanocrystals were then simulated by Blochwave and multislice simulations to determine the atomic
Wei-Qiang Han et al.
Journal of the American Chemical Society, 127(37), 12814-12815 (2005-09-15)
Well-crystalline CeO(2-x) nanotubes are synthesized via a mild hydrothermal reaction route using cerium nitrate and ammonium hydroxide as reactants. The CeO(2-x) nanotubes have the same structure as the bulk CeO2 but larger lattice parameter. The measurement of the ratio of

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service