Skip to Content
Merck
All Photos(1)

Key Documents

324108

Sigma-Aldrich

Trimethylphosphine solution

1.0 M in THF

Synonym(s):

PMe3, Trimethylphosphorus

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3P
CAS Number:
Molecular Weight:
76.08
Beilstein:
969138
MDL number:
UNSPSC Code:
12352128
PubChem Substance ID:
NACRES:
NA.22

form

liquid

reaction suitability

reagent type: ligand
reaction type: Mitsunobu Reaction

reagent type: ligand
reaction type: Wittig Reaction

concentration

1.0 M in THF

density

0.872 g/mL at 25 °C

functional group

phosphine

SMILES string

CP(C)C

InChI

1S/C3H9P/c1-4(2)3/h1-3H3

InChI key

YWWDBCBWQNCYNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trimethylphosphine is the reagent used in Mitsunobu reaction. It participates in the transformation of azides into carbamates, aziridines from azidoalcohols, iminophosphoranes and aza-Wittig reaction. Electron diffraction study of trimethylphosphine has been reported.

Application

Trimethylphosphine (PMe3) solution is the suitable reagent used in the synthesis of Fe/Te cluster type complex, Fe6Te8(PMe3)6. It may be employed as a probe to investigate the acid sites in Y-zeolite. It may be used for the synthesis of hexakis(trimethylphosphine)tris-μ-methylene-diruthenium(III).

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-22.0 °F - closed cup

Flash Point(C)

-30 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acid sites in zeolite Y: a solid-state NMR and infrared study using trimethylphosphine as a probe molecule.
Lunsford JH, et al.
Journal of the American Chemical Society, 107(6), 1540-1547 (1985)
Effect of Diverse Ligands on the Course of a Molecules-to-Solids Process and Properties of Its Intermediates.
Steigerwald ML, et al.
Inorganic Chemistry, 33(15), 3389-3395 (1994)
Electron diffraction study of the structure of trimethylphosphine.
Bartell LS and Brockway LO.
J. Chem. Phys. , 32(2), 512-515 (1960)
Synthesis and x-ray crystal structures of hexakis (trimethylphosphine) tris-. mu.-methylene-diruthenium (III) and its mono-and dicationic derivatives.
Hursthouse MB, et al.
Journal of the American Chemical Society, 101(15), 4128-4139 (1979)
William Levason et al.
Dalton transactions (Cambridge, England : 2003), 43(25), 9557-9566 (2014-05-16)
The reactions of the soft diphosphines o-C6H4(PMe2)2, Me2P(CH2)2PMe2, Et2P(CH2)2PEt2 or o-C6H4(PPh2)2 with NbF5 or TaF5 in anhydrous MeCN solution produce [MF4(diphosphine)2][MF6] (M = Nb or Ta), which have been characterised by microanalysis, IR, (1)H, (19)F{(1)H}, (31)P{(1)H} and (93)Nb NMR spectroscopy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service