Skip to Content
Merck
All Photos(1)

Documents

T84603

Sigma-Aldrich

Triphenylphosphine oxide

98%

Synonym(s):

Ph3PO, TPPO, Triphenyl phosphorus oxide, Triphenylphosphine monoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3PO
CAS Number:
Molecular Weight:
278.28
Beilstein:
745854
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

reaction suitability

reagent type: ligand
reaction type: Coupling Reactions

reagent type: ligand
reaction type: Epoxidations

reagent type: ligand
reaction type: Michael Reaction

mp

150-157 °C (lit.)

functional group

phosphine oxide

SMILES string

O=P(c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C18H15OP/c19-20(16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H

InChI key

FIQMHBFVRAXMOP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Triphenylphosphine oxide can be used:
  • As a catalyst in Appel-type chlorination reaction of acyclic primary and secondary alcohols.
  • As a catalyst in stereoselective poly and dibromination of α,β-unsaturated esters and β,γ-unsaturated α-ketoester compounds.
  • As a promotor in the diastereoselective synthesis of α-ribofuranosides through ribofuranosylation of alcohols with ribofuranosyl iodides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

356.0 °F

Flash Point(C)

180 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Triphenylphosphine oxide-catalyzed stereoselective poly-and dibromination of unsaturated compounds
Yu T, et al.
Chemical Communications (Cambridge, England), 50(58), 7817-7820 (2014)
Phosphine oxide-catalysed chlorination reactions of alcohols under Appel conditions
Denton RM, et al.
Chemical Communications (Cambridge, England), 46(17), 3025-3027 (2010)
?-Selective Ribofuranosylation of Alcohols with Ribofuranosyl Iodides and Triphenylphosphine Oxide
Oka N, et al.
The Journal of Organic Chemistry, 79(16), 7656-7664 (2014)
Ya-Kun Wang et al.
Nature communications, 11(1), 3674-3674 (2020-07-24)
Metal halide perovskites have emerged as promising candidates for solution-processed blue light-emitting diodes (LEDs). However, halide phase segregation - and the resultant spectral shift - at LED operating voltages hinders their application. Here we report true-blue LEDs employing quasi-two-dimensional cesium
Stephen M Richoll et al.
Journal of chromatography. A, 1127(1-2), 147-153 (2006-07-04)
A versatile procedure has been developed and validated for the determination of triphenylphosphine oxide (TPPO) at low levels in various active pharmaceutical ingredients (APIs). This procedure incorporates the use of the novel hollow-fiber liquid-phase microextraction (LPME) for the measurement of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service