Skip to Content
Merck
All Photos(1)

Documents

245275

Sigma-Aldrich

Triethylphosphine

99%

Synonym(s):

Triethylphosphorus

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3P
CAS Number:
Molecular Weight:
118.16
Beilstein:
969170
EC Number:
MDL number:
UNSPSC Code:
12352128
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

reaction suitability

reagent type: ligand
reaction type: Arylations

refractive index

n20/D 1.456 (lit.)

bp

127-128 °C (lit.)

density

0.802 g/mL at 20 °C (lit.)

functional group

phosphine

SMILES string

CCP(CC)CC

InChI

1S/C6H15P/c1-4-7(5-2)6-3/h4-6H2,1-3H3

InChI key

RXJKFRMDXUJTEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Triethylphosphine is generally used as a ligand in the organometallic chemistry. It can be used in:
  • Synthesis of the tetrahedrally coordinated L3Fe-Nx complex with a terminal nitride group.
  • Synthesis of (PEt3)2Ni(η2-C14H10), a source of the reactive Ni(PEt3)2 moiety.
  • Synthesis of dinuclear rhodium complexes with triethyllphosphane bridges.

Other Notes

This container should be opened only by a technically qualified person.

related product

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Pyr. Liq. 1 - Skin Corr. 1B

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Breaking the Rule: Synthesis and Molecular Structure of Dinuclear Rhodium Complexes with Bridging and Semibridging Trialkylphosphane Ligands.
Pechmann T, et al.
Angewandte Chemie (International Edition in English), 39(21), 3909-3911 (2000)
Unexpected Intermediates and Products in the C? F Bond Activation of Tetrafluorobenzenes with a Bis (triethylphosphine) Nickel Synthon: Direct Evidence of a Rapid and Reversible C? H Bond Activation by Ni (0).
Johnson S A, et al.
Journal of the American Chemical Society, 130(51), 17278-17280 (2008)
A Tetrahedrally Coordinated L3Fe? N x Platform that Accommodates Terminal Nitride (FeIV? N) and Dinitrogen (FeI? N2? FeI) Ligands.
Betley T A and Peters J C
Journal of the American Chemical Society, 126(20), 6252-6254 (2004)
Quintin R Sheridan et al.
The journal of physical chemistry. B, 120(46), 11951-11960 (2016-10-27)
A combination of X-ray scattering experiments and molecular dynamics simulations were conducted to investigate the structure of ionic liquids (ILs) which chemically bind CO
Matthew R Richardson et al.
Proteomics, 9(9), 2468-2483 (2009-04-30)
Although low-density lipoprotein (LDL) plays a predominant role in atherogenesis, the low-density lipoproteome has not been fully characterized. Moreover, alterations from a Western diet, diabetes, and physical inactivity on this proteome have yet to be determined. Accordingly, relative quantification was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service