Skip to Content
Merck
All Photos(1)

Key Documents

298190

Sigma-Aldrich

Cerium(III) chloride

AnhydroBeads, −10 mesh, 99.9%

Synonym(s):

Cerium trichloride, Cerous chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CeCl3
CAS Number:
Molecular Weight:
246.48
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.9%

product line

AnhydroBeads

Quality Level

Assay

99.9%

reaction suitability

reagent type: catalyst
core: cerium

impurities

≤1500.0 ppm Trace Rare Earth Analysis

particle size

−10 mesh

mp

848 °C (lit.)

density

3.97 g/mL at 25 °C (lit.)

SMILES string

Cl[Ce](Cl)Cl

InChI

1S/Ce.3ClH/h;3*1H/q+3;;;/p-3

InChI key

VYLVYHXQOHJDJL-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

When added to nitrile aldol reactions, diastereoselectivity is enhanced, providing high yields of β-hydroxynitriles.

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Schneider et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(50), 12243-12252 (2017-03-24)
A series of heteroleptic tris(cyclopentadienyl) Ce
Xiao, Z. Timberlake, J.W.
Tetrahedron, 54, 4211-4211 (1998)
Zhe Cheng et al.
Environmental toxicology, 27(12), 707-718 (2011-03-09)
Experimental studies have demonstrated that lanthanides could impair cognitive functions of children and animals, but very little is known about the hippocampal apoptosis and its molecular mechanism. The study investigated the signal pathway of hippocampal apoptosis induced by intragastric administration
Florian J Wegehaupt et al.
Archives of oral biology, 55(6), 441-446 (2010-04-20)
To investigate the effect of cerium chloride, cerium chloride/fluoride and fluoride application on calcium release during erosion of treated dentine. Forty dentine samples were prepared from human premolars and randomly assigned to four groups (1-4). Samples were treated twice a
Masami Kawagoe et al.
Journal of trace elements in medicine and biology : organ of the Society for Minerals and Trace Elements (GMS), 22(1), 59-65 (2008-03-06)
We evaluated tissue changes associated with cerium chloride administration via gavage to adult mice, via milk to neonatal mice and transplacentally to fetal mice. Change in adults consisted of extensive pulmonary hemorrhage, pulmonary venous congestion, thickened alveolar septae, hepatic necrosis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service