Skip to Content
Merck
All Photos(4)

Key Documents

228931

Sigma-Aldrich

Cerium(III) chloride heptahydrate

99.9% trace metals basis

Synonym(s):

Cerous chloride heptahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CeCl3 · 7H2O
CAS Number:
Molecular Weight:
372.58
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.9% trace metals basis
form:
crystals and lumps

Quality Level

Assay

99.9% trace metals basis

form

crystals and lumps

reaction suitability

reagent type: catalyst
core: cerium

impurities

≤1500.0 ppm Trace Rare Earth Analysis

density

~3.94 g/mL at 25 °C (lit.)

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].Cl[Ce](Cl)Cl

InChI

1S/Ce.3ClH.7H2O/h;3*1H;7*1H2/q+3;;;;;;;;;;/p-3

InChI key

KPZSTOVTJYRDIO-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Cerium(III) chloride heptahydrate is a hygroscopic crystalline solid. It is soluble in water and forms acidic solutions. It is used as a cerium source in various applications such as nanometal oxide synthesis and catalysis.

Application

Cerium(III) chloride heptahydrate can be used:
  • As a precursor to prepare cerium oxide nanoparticles for biomedical applications and photocatalytic degradation.
  • As a solution to fabricate thin films of CeO2 on glass substrates by the spray pyrolysis process.
  • As a dopant to fabricate ZnO and CeO2 nanocrystals for electrochemical sensing of H2O2 and photocatalytic degradation of Rhodamine B and Congo red dyes.
  • As an additive to prepare corrosion-inhibiting formulations and coatings.
  • To synthesize carbon nanofiber composites to fabricate high-temperature polymer electrolyte membrane fuel cell cathodes.
  • As a support for the combination of cerium(III)chloride heptahydrate and sodium iodide supported on silica gel to promoteMichael-type additions. These catalysts are used to convert from indolesand nitroalkenes to 2-indolyl-1-nitroalkane derivatives in good yields.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Chem. Res. (M), 668-668 (2006)
Tetrahedron, 63, 892-892 (2007)
Zhe Cheng et al.
Environmental toxicology, 27(12), 707-718 (2011-03-09)
Experimental studies have demonstrated that lanthanides could impair cognitive functions of children and animals, but very little is known about the hippocampal apoptosis and its molecular mechanism. The study investigated the signal pathway of hippocampal apoptosis induced by intragastric administration
Florian J Wegehaupt et al.
Archives of oral biology, 55(6), 441-446 (2010-04-20)
To investigate the effect of cerium chloride, cerium chloride/fluoride and fluoride application on calcium release during erosion of treated dentine. Forty dentine samples were prepared from human premolars and randomly assigned to four groups (1-4). Samples were treated twice a
Xiaoming Tao et al.
Organic letters, 15(1), 72-75 (2012-12-25)
Dynamic kinetic resolution of various α-amido-β-keto phosphonates via asymmetric hydrogenation proceeded efficiently to give the corresponding β-hydroxy-α-amido phosphonates in high diastereo- and enantioselectivities (up to 99:1 syn/anti, 99.8% ee). The addition of catalytic amounts of CeCl(3)·7H(2)O is necessary to achieve

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service