Skip to Content
Merck
All Photos(1)

Key Documents

295051

Sigma-Aldrich

1-Butene

≥99%

Synonym(s):

α-Butene, α-Butylene, 2-Butylene, ethylenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2CH3
CAS Number:
Molecular Weight:
56.11
Beilstein:
1098262
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

1.93 (vs air)

vapor pressure

1939 mmHg ( 21.1 °C)

Assay

≥99%

form

gas

autoignition temp.

723 °F

expl. lim.

9.3 %

bp

−6.3 °C (lit.)

mp

−185 °C (lit.)

functional group

allyl

SMILES string

CCC=C

InChI

1S/C4H8/c1-3-4-2/h3H,1,4H2,2H3

InChI key

VXNZUUAINFGPBY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

  • Development and characterization of ZnO, Au/ZnO and Pd/ZnO thin films through their adsorptive and catalytic properties.: This paper discusses the synthesis and catalytic applications of ZnO-based thin films, where 1-butene serves as a probe molecule to study adsorption and catalysis, contributing to advancements in material science and catalysis technology (Giannoudakos et al., 2008).

Packaging

Supplied in a Sure/Pac cylinder and has a brass needle valve with a male 1/4" NPTF outlet thread installed. Before using the cylinder, ensure that the valve is closed, then remove the galvanized steel hex cap that seals the outlet valve.

Compatible with the following:

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

also commonly purchased with this product

Product No.
Description
Pricing

hose barb

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1A - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

-112.0 °F - closed cup

Flash Point(C)

-80 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emilio Marengo et al.
Talanta, 77(3), 1111-1119 (2008-12-10)
This paper reports the development of calibration models for quality control in the production of ethylene/propylene/1-butene terpolymers by the use of multivariate tools and FT-IR spectroscopy. 1-Butene concentration prediction is achieved in terpolymers by coupling FT-IR spectroscopy to multivariate regression
Lei Li et al.
Macromolecular rapid communications, 32(22), 1834-1838 (2011-09-20)
The deformation of isotactic poly-1-butene (iPB-1) matrix, during the CO(2) -assisted foaming process, makes the iPB-1 melt crystallize into form I rather than form I', which crystallizes after annealing under high-pressure CO(2) without foaming. The result provides a new strategy
Chia-Hsiang Lai et al.
Environmental monitoring and assessment, 184(7), 4015-4028 (2011-08-09)
The concentrations of 56 volatile organic hydrocarbons (VOCs) were measured simultaneously in the southbound bore, the northbound bore and the exhaust air shafts of the Hsuehshan tunnel near Yilan, Taiwan during 2007 and 2008. A total of 60 integrated air
Fang Wu et al.
The journal of physical chemistry. A, 112(18), 4360-4366 (2008-05-02)
The valence-shell electron momentum distributions for 1-butene are measured by electron momentum spectroscopy (EMS) employing non-coplanar symmetric geometry. The experimental electron momentum distributions are compared with the density functional theory (DFT) calculations using different-sized basis sets. Although the two conformers
Johanna L Miller et al.
The journal of physical chemistry. A, 109(18), 4038-4048 (2006-07-13)
The work presented here is the first in a series of studies that use a molecular beam scattering technique to investigate the unimolecular reaction dynamics of C(4)H(7) radical isomers. Photodissociation of the halogenated precursor 2-bromo-1-butene at 193 nm under collisionless

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. In which type of packaging would this product be supplied?

    1 answer
    1. This product is supplied in a Sure/Pac™ cylinder. For more information please see the technical bulletin: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/393/908/al_techbull_al136.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service