Skip to Content
Merck
All Photos(1)

Key Documents

295086

Sigma-Aldrich

trans-2-Butene

≥99%

Synonym(s):

β-trans-Butylene, (2E)-2-Butene, (E)-2-Butene, 2-trans-Butene, 2-trans-Butylene, E-Butene, trans-1,2-Dimethylethylene, trans-2-Butylene, trans-Butene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCH3
CAS Number:
Molecular Weight:
56.11
Beilstein:
1718756
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

2 (vs air)

Quality Level

vapor pressure

2575 mmHg ( 37.7 °C)

Assay

≥99%

autoignition temp.

615 °F

expl. lim.

9.7 %

bp

1 °C (lit.)

mp

−105 °C (lit.)

SMILES string

CC=CC

InChI

1S/C4H8/c1-3-4-2/h3-4H,1-2H3/b4-3+

InChI key

IAQRGUVFOMOMEM-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

trans-2-Butene can be used as a monomer unit to produce high molecular weight polyolefins with well-defined microstructures by nickle-catalyzed polymerization reaction. It is also used as a reactant in the isomerization reaction to produce the corresponding isomer using acid-activated catalysts.[1][2]

Packaging

Supplied in a Sure/Pac cylinder and has a brass needle valve with a male 1/4" NPTF outlet thread installed. Before using the cylinder, ensure that the valve is closed, then remove the galvanized steel hex cap that seals the outlet valve.

Compatible with the following:

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

also commonly purchased with this product

Product No.
Description
Pricing

hose barb

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1 - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Ni (II)-catalyzed polymerization of trans-2-butene
Leatherman MD and Brookhart M
Macromolecules, 34(9), 2748-2750 (2001)
Isomerization of cis-2-butene and trans-2-butene catalyzed by acid-and ion-exchanged smectite-type clays
Moronta A, et al.
Applied Clay Science, 29(2), 117-123 (2005)
Duangkamol Gleeson
Journal of computer-aided molecular design, 22(8), 579-585 (2008-03-18)
In this study the results from a series of calculations are reported that probe the influence of the QM cluster size and the extended framework treatment in ONIOM calculations. This is done by comparing the differences in the structures and
Philip T M Carlsson et al.
Physical chemistry chemical physics : PCCP, 14(45), 15637-15640 (2012-10-24)
Recent studies have suggested that the reaction of stabilised Criegee Intermediates (CIs) with sulfur dioxide (SO(2)), leading to the formation of a carbonyl compound and sulfur trioxide, is a relevant atmospheric source of sulfuric acid. Here, the significance of this
Franz Worek et al.
Archives of toxicology, 86(9), 1379-1386 (2012-03-23)
The reactivation of organophosphorus compound (OP)-inhibited acetylcholinesterase (AChE) by oximes is inadequate in case of different OP nerve agents. This fact led to the synthesis of numerous novel oximes by different research groups in order to identify more effective reactivators.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service