Skip to Content
Merck
All Photos(1)

Documents

241997

Sigma-Aldrich

1-Pentene

98%

Synonym(s):

α-n-Amylene, 1-Methyl-3-butene, 4-Methyl-1-butene, Propylethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH=CH2
CAS Number:
Molecular Weight:
70.13
Beilstein:
1731629
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.4 (vs air)

vapor pressure

10.27 psi ( 20 °C)

Assay

98%

form

liquid

autoignition temp.

522 °F

expl. lim.

8.7 %

refractive index

n20/D 1.371 (lit.)

bp

29.9-30.1 °C (lit.)

solubility

alcohol: miscible(lit.)
benzene: miscible(lit.)
diethyl ether: miscible(lit.)
water: insoluble(lit.)

density

0.641 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCC=C

InChI

1S/C5H10/c1-3-5-4-2/h3H,1,4-5H2,2H3

InChI key

YWAKXRMUMFPDSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The bacterial mutagenecity of 1-pentene was studied.

Application

1-Pentene was used to develop a rhodium/xantphos homogeneous catalyst system for direct chemo- and regioselective mono-N-alkylation of primary amides with 1-alkenes.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 1 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-59.8 °F - closed cup

Flash Point(C)

-51 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saeed Raoufmoghaddam et al.
ChemSusChem, 6(9), 1759-1773 (2013-09-07)
A rhodium/xantphos homogeneous catalyst system has been developed for direct chemo- and regioselective mono-N-alkylation of primary amides with 1-alkenes and syngas through catalytic hydroamidomethylation with 1-pentene and acetamide as model substrates. For appropriate catalyst performance, it appears to be essential
Götz A Westphal et al.
BioMed research international, 2014, 592434-592434 (2014-02-11)
Amylenes are unsaturated hydrocarbons (C5H10), such as 1-pentene, 2-pentene, 2-methyl-but-1-en (3-methyl-1-butene), 2-methyl-but-2-en (isopentene), and 3-methyl-but-1-en. We investigated bacterial mutagenicity of 1-pentene, 2-pentene, and 3-methyl-but-1-en in the Ames test. 2-Pentene was investigated as racemate and as pure diastereomers. We included the
S R Merrigan et al.
Organic letters, 1(2), 327-329 (2000-05-24)
[formula: see text] The 13C and 2H kinetic isotope effects for the bromination of 1-pentene with Br2 in CCl4 were determined and interpreted with the aid of calculationally predicted isotope effects. The isotope effects observed are consistent with rate-limiting bromonium
Yu Shan Susanna Wan et al.
Chemical communications (Cambridge, England), (8)(8), 878-879 (2002-07-19)
A zeolite-based microengineered reactor was fabricated and tested for 1-pentene epoxidation over titanium silicalite-1 (TS-1) catalyst, which has been selectively incorporated within the microreactor channel using a new synthesis procedure.
Chen Zhao et al.
Urology, 82(3), 744-744 (2013-07-31)
To analyze the gas generated from the transurethral resection of the prostate (TURP) and transurethral resection of bladder (TURB) tumor. Thirty-six smoke samples were collected from a continuous irrigation suction system during the TURP and the TURB. Then, they were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service