294454
Potassium formate
ReagentPlus®, 99%
Synonym(s):
Formic acid potassium salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HCOOK
CAS Number:
Molecular Weight:
84.12
Beilstein:
3623272
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22
Assay:
99%
form:
solid
Recommended Products
Quality Level
product line
ReagentPlus®
Assay
99%
form
solid
mp
165-168 °C (lit.)
SMILES string
[K+].[O-]C=O
InChI
1S/CH2O2.K/c2-1-3;/h1H,(H,2,3);/q;+1/p-1
InChI key
WFIZEGIEIOHZCP-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Application
Potassium formate has been used as a biochemical component for the synthesis of liquid phantom to study the Raman reconstruction by Wiener estimation.
It can be used:
It can be used:
- For the hydroxycarbonylation of aryl halides in the presence of 1–5 mol % of CO.
- As an additive for thermal activation of sodium peroxydisulfate.
- As a reducing agent to suppress the undesired boronic acid homocoupling reaction in Suzuki cross-coupling reaction.
Legal Information
ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Adrianna Nogalska et al.
Membranes, 10(11) (2020-11-25)
Membrane electrode assemblies (MEAs) with palladium catalysts were successfully prepared by using a home-made manual pressing system with Nafion glue application that contributed to a decrease of additional energy consumption. The catalyst coated membranes were prepared with supported palladium on
Suppression of a palladium-mediated homocoupling in a Suzuki cross-coupling reaction. Development of an impurity control strategy supporting synthesis of LY451395.
Miller WD, et al.
Organic Process Research & Development, 11(3), 359-364 (2007)
A Method to Create a Universal Calibration Dataset for Raman Reconstruction Based on Wiener Estimation.
Chen S, et al.
IEEE Journal of Selected Topics in Quantum Electronics, 22(3), 1-7 (2016)
Effective palladium-catalyzed hydroxycarbonylation of aryl halides with substoichiometric carbon monoxide.
Korsager S, et al.
Journal of the American Chemical Society, 135(8), 2891-2894 (2013)
Verónica C Mora et al.
Chemosphere, 75(10), 1405-1409 (2009-03-17)
The kinetics and mechanism of the thermal activation of peroxydisulfate, in the temperature range from 60 to 80 degrees C, was investigated in the presence and absence of sodium formate as an additive to turn the oxidizing capacity of the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service