Skip to Content
Merck
All Photos(3)

Documents

M29800

Sigma-Aldrich

2-Methylbenzimidazole

98%

Synonym(s):

2-Methyl-1H-1,3-benzodiazole, 2-Methyl-1H-benzimidazole, 2-Methyl-1H-benzo[d]imidazole, 2-Methylbenzoimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8N2
CAS Number:
Molecular Weight:
132.16
Beilstein:
112264
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
32151902
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

175-177 °C (lit.)

SMILES string

Cc1nc2ccccc2[nH]1

InChI

1S/C8H8N2/c1-6-9-7-4-2-3-5-8(7)10-6/h2-5H,1H3,(H,9,10)

InChI key

LDZYRENCLPUXAX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • 2-Methylbenzimidazole is an important pharmacophore widely used in medicinal chemistry for the synthesis of various antibacterial and antifungal agents.
  • It can be used as a key precursor to synthesize substituted benzimidazo[1,2-a]quinolones.
  • It can be used in the synthesis of reversible solid-to-liquid phase transition coordination polymer crystals.
  • 2-Methylbenzimidazole also exhibits corrosion inhibition.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reversible solid-to-liquid phase transition of coordination polymer crystals.
Umeyama D, et al.
Journal of the American Chemical Society, 137(2), 864-870 (2015)
Theoretical study of benzimidazole and its derivatives and their potential activity as corrosion inhibitors.
Obot I B and Obi-Egbedi N O
Corrosion Science, 52(2), 657-660 (2010)
Novel strategy for synthesis of substituted benzimidazo [1, 2-a] quinolines.
Kato J Y, et al.
Organic Letters, 15(14), 3794-3797 (2013)
Synthesis and Antimicrobial Activity of Some Benzimidazole and 2-Methylbenzimidazole Derivatives.
Jain P and Tiwari M
Asian Journal of Chemistry, 29(4), 838-838 (2017)
Synthesis and biological evaluation of substituted benzimidazoles.
Shah K, et al.
Journal of the Indian Chemical Society, 93, 1009-1018 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service