Skip to Content
Merck
All Photos(1)

Key Documents

433764

Sigma-Aldrich

Cesium formate

98%

Synonym(s):

Formic acid cesium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOOCs
CAS Number:
Molecular Weight:
177.92
Beilstein:
3912426
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
crystals and lumps

grade

for analytical purposes

Assay

98%

form

crystals and lumps

SMILES string

[Cs+].[H]C([O-])=O

InChI

1S/CH2O2.Cs/c2-1-3;/h1H,(H,2,3);/q;+1/p-1

InChI key

ATZQZZAXOPPAAQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT RE 2 Oral - STOT SE 2 Oral

Target Organs

Nervous system, Nervous system,Blood

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Grant T Corbett et al.
The Journal of biological chemistry, 288(12), 8299-8312 (2013-02-14)
Neurotrophins, such as brain-derived neurotrophic factor (BDNF) and neurotrophin-3 (NT-3), are believed to be genuine molecular mediators of neuronal growth and homeostatic synapse activity. However, levels of these neurotrophic factors decrease in different brain regions of patients with Alzheimer disease
Lydia Beyer et al.
Journal of bacteriology, 195(7), 1428-1435 (2013-01-22)
Enterobacteria such as Escherichia coli generate formate, lactate, acetate, and succinate as major acidic fermentation products. Accumulation of these products in the cytoplasm would lead to uncoupling of the membrane potential, and therefore they must be either metabolized rapidly or
Sai Duan et al.
Physical chemistry chemical physics : PCCP, 15(13), 4625-4633 (2013-02-21)
Local structures and adsorption energies of a formic acid molecule and its decomposed intermediates (H, O, OH, CO, HCOO, and COOH) on highly electrocatalytically active mushroom-like Au-core@Pd-shell@Pt-cluster nanoparticles with two atomic layers of the Pd shell and stoichiometric Pt coverage
Pavel Kubáň et al.
Analytica chimica acta, 768, 82-89 (2013-03-12)
A simple method using direct injection of human blood samples and quantitative analysis of formate was developed for rapid diagnosis of methanol poisoning. A sample pretreatment device including a 500Da molecular weight cut-off dialysis membrane was in-line coupled to capillary
J K Htoo et al.
Journal of animal science, 90 Suppl 4, 346-349 (2013-02-13)
Dietary inclusion of salts of organic acids may modulate intestinal microbiota and enhance performance of pigs. Published data on the effects of potassium formate (KF; pH 6.0-8.5) on pig performance are scarce compared with that of potassium diformate (KDF; pH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service