Skip to Content
Merck
All Photos(1)

Key Documents

294047

Sigma-Aldrich

cis-2,3-Epoxybutane

97%

Synonym(s):

cis-β-Butylene oxide, cis-2,3-Dimethyloxirane, cis-2-Butene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
Beilstein:
79770
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

refractive index

n20/D 1.383 (lit.)

bp

60-61 °C (lit.)

mp

−84-−83 °C (lit.)

density

0.826 g/mL at 25 °C (lit.)

SMILES string

C[C@@H]1O[C@@H]1C

InChI

1S/C4H8O/c1-3-4(2)5-3/h3-4H,1-2H3/t3-,4+

InChI key

PQXKWPLDPFFDJP-ZXZARUISSA-N

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emily C Minerath et al.
Environmental science & technology, 43(5), 1386-1392 (2009-04-09)
Polyols and sulfate esters have recently been identified in the secondary organic aerosol (SOA) formed in the photooxidation of biogenic hydrocarbons both in the laboratory and under ambient atmospheric conditions. In the present study, the potential role of the reactions
C de Meester
Toxicology letters, 21(3), 255-262 (1984-06-01)
The mutagenicity of 2,3- epoxybutane (2,3-EB) towards Salmonella FFphimurium TA1530 is increased by rat liver S9 mix. This enhancement can be induced by Aroclor 1254 (ARO) or phenobarbitone (PB) pretreatment. Comparative assays with purified subcellular liver fractions demonstrated that cytosol
C de Meester et al.
Journal of applied toxicology : JAT, 2(6), 284-288 (1982-12-01)
The compound 2,3-epoxybutane (2,3-EB) is a direct mutagen for the base-pair-substitution-sensitive strains TA 1530, TA 1535 and TA 100, in the absence and in the presence of metabolic activation by liver S9-mix. Under the same experimental conditions, the 2-methyl derivative
P Castelain et al.
Mutagenesis, 8(5), 387-393 (1993-09-01)
Four structurally related aliphatic epoxides (1,2-epoxypropane, 1,2-epoxyisobutane, cis- and trans-2,3-epoxybutane) have been tested in the Salmonella/microsome assay, modified for volatile substances, using the strains TA1535 and TA100. The aim of the study was to evaluate the effect of methylation on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service