531537
1,2-Epoxy-2-methylpropane
97%
Synonym(s):
1,2-Epoxy-isobutane, 2,2-Dimethyloxirane, Isobutylene oxide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Quality Level
Assay
97%
refractive index
n20/D 1.374 (lit.)
bp
50-51 °C (lit.)
density
0.812 g/mL at 25 °C (lit.)
SMILES string
CC1(C)CO1
InChI
1S/C4H8O/c1-4(2)3-5-4/h3H2,1-2H3
InChI key
GELKGHVAFRCJNA-UHFFFAOYSA-N
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - Flam. Liq. 2 - Muta. 2 - Skin Corr. 1C
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
-26.5 °F
Flash Point(C)
-32.5 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
M Cornet et al.
Archives of toxicology, 65(4), 263-267 (1991-01-01)
Until now, no data are available concerning the biotransformation and toxicity of 2-methylpropene (or isobutene), a gaseous alkene widely used in industry (rubber, fuel additives, plastic polymers, adhesives, antioxidants). In this work, the biotransformation of 2-methylpropene (MP) has been studied
U Jorritsma et al.
Mutagenesis, 10(2), 101-104 (1995-03-01)
2-Methylpropene (isobutene), a gaseous compound widely used in chemical industries, is metabolized to the epoxide 2-methyl-1,2-epoxypropane. The parent compound has previously been shown to be non-mutagenic in a modified Ames test, whereas the epoxide metabolite gave a positive result. In
Paul R Carlier et al.
The Journal of organic chemistry, 71(4), 1592-1597 (2006-02-14)
Protonated epoxides feature prominently in organic chemistry as reactive intermediates. Herein, we describe 10 protonated epoxides using B3LYP, MP2, and CCSD/6-311++G calculations. Relative to CCSD, B3LYP consistently overestimates the C2-O bond length. Protonated 2-methyl-1,2-epoxypropane is the most problematic species studied
M Cornet et al.
Chemical research in toxicology, 8(7), 987-992 (1995-10-01)
The biotransformation of 2-methylpropene, a gaseous alkene widely used in industry, was investigated in vitro in liver tissue of rats, mice, and humans. Interspecies comparison revealed that the lowest levels of the primary epoxide metabolite were detected in incubations of
M Cornet et al.
Mutation research, 271(3), 213-221 (1992-06-01)
The mutagenic properties of 2-methylpropene (MP) and 2-methyl-1,2- epoxypropane (MEP) were investigated in the Salmonella assay. A simple exposure system, consisting of gastight tissue culture flasks, was used. This method has the advantage that the volatile test chemical is present
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service