127574
3,4-Epoxy-1-butene
98%
Synonym(s):
2-Vinyloxirane, 3,4-Epoxy-1-butene, Butadiene monoxide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C4H6O
CAS Number:
Molecular Weight:
70.09
Beilstein:
103170
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Quality Level
Assay
98%
form
liquid
refractive index
n20/D 1.417 (lit.)
bp
65-66 °C (lit.)
density
0.87 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
C=CC1CO1
InChI
1S/C4H6O/c1-2-4-3-5-4/h2,4H,1,3H2
InChI key
GXBYFVGCMPJVJX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
accessory
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
-58.0 °F - closed cup
Flash Point(C)
-50 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Eduardo Cemeli et al.
Mutation research, 664(1-2), 69-76 (2009-05-12)
The toxicity of butadiene and styrene is exerted by their metabolites. Such metabolites have been extensively scrutinized at the in vitro level demonstrating evident genotoxic properties. In monitoring, a diverse range of outcomes has been produced. Additionally, epidemiological studies in
Stereoselective ring expansion of vinyl oxiranes: mechanistic insights and natural product total synthesis.
Matthew Brichacek et al.
Angewandte Chemie (International ed. in English), 49(9), 1648-1651 (2010-02-06)
L Recio et al.
Chemico-biological interactions, 135-136, 325-341 (2001-06-09)
1,3-Butadiene (BD) is a multisite carcinogen and is mutagenic in multiple tissues of B6C3F1 mice. BD is bioactivated to at least three directly mutagenic metabolites: 1,2-epoxybutene (EB), 1,2-epoxy-3,4-butanediol (EBD), and 1,2,3,4-diepoxybutane (DEB). However, the contribution of these individual metabolites to
Some insights into the mode of action of butadiene by examining the genotoxicity of its metabolites.
A D Kligerman et al.
Chemico-biological interactions, 166(1-3), 132-139 (2006-05-16)
1,3-Butadiene (BTD) is an important commodity chemical and air pollutant that has been shown to be a potent carcinogen in mice, and to a lesser extent, a carcinogen in rats. To better assess butadiene's carcinogenic risk to humans, it is
Charlotta Fred et al.
Mutation research, 585(1-2), 21-32 (2005-06-01)
1,3-Butadiene and isoprene (2-methyl-1,3-butadiene) are chemically related substances that are carcinogenic to rodents. The overall aim of this work is to elucidate the role of the genotoxic action of diepoxide metabolites in the carcinogenesis of the dialkenes. In vivo doses
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service