Skip to Content
Merck
All Photos(2)

Documents

247502

Sigma-Aldrich

Cumene hydroperoxide

technical grade, 80%

Synonym(s):

α,α-Dimethylbenzyl hydroperoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C(CH3)2OOH
CAS Number:
Molecular Weight:
152.19
Beilstein:
1908117
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

vapor density

5.4 (vs air)

vapor pressure

<0.03 mmHg ( 20 °C)

Assay

80%

form

liquid

reaction suitability

reagent type: oxidant

refractive index

n20/D 1.5210

bp

100-101 °C/8 mmHg (lit.)

solubility

ethanol: 1 mL/mL, clear, colorless to very faintly yellow

density

1.03 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC(C)(OO)c1ccccc1

InChI

1S/C9H12O2/c1-9(2,11-10)8-6-4-3-5-7-8/h3-7,10H,1-2H3

InChI key

YQHLDYVWEZKEOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Epoxidation reagent for allylic alcohols and fatty acid esters, as an initiator for radical polymerization.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 1B - Eye Dam. 1 - Org. Perox. F - Skin Corr. 1B - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Lungs, Respiratory system

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

174.2 °F

Flash Point(C)

79 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Gulfam Ahmad et al.
Antioxidants (Basel, Switzerland), 8(9) (2019-09-22)
Activated neutrophils release myeloperoxidase that produces the potent oxidant hypochlorous acid (HOCl). Exposure of the oxygen transport protein horse heart myoglobin (hhMb) to HOCl inhibits Iron III (Fe(III))-heme reduction by cytochrome b5 to oxygen-binding Iron II (Fe(II))Mb. Pathological concentrations of
Maren C Podszun et al.
Redox biology, 37, 101710-101710 (2020-09-14)
Oxidative stress (OS) in non-alcoholic fatty liver disease (NAFLD) promotes liver injury and inflammation. Treatment with vitamin E (α-tocopherol, αT), a lipid-soluble antioxidant, improves liver injury but also decreases steatosis, thought to be upstream of OS, through an unknown mechanism.
D Xia et al.
Cell motility and the cytoskeleton, 26(2), 115-124 (1993-01-01)
We used a cell free system Bouché et al.: J. Cell Biol. 107:587-596, 1988] to study the incorporation of actin into myofibrils. We used alpha-skeletal muscle actin and actins with substitutions of either His73 [Solomon and Rubenstein: J. Biol.Chem. 262:11382
Takaya Nakazawa et al.
The Journal of steroid biochemistry and molecular biology, 169, 69-76 (2016-03-10)
24(S)-Hydroxycholesterol (24S-OHC) has diverse physiological and pathological functions. In particular, cytotoxic effects of 24S-OHC in neuronal cells are important in development of neurodegenerative diseases. 24S-OHC induces necroptosis-like cell death in SH-SY5Y cells expressing little caspase-8. In the present study, 24S-OHC
Chao Wang et al.
Molecular nutrition & food research, 62(18), e1700840-e1700840 (2018-02-01)
This study aims to investigate the role of the epigenetic regulator SET domain-containing lysine methyltransferase 7 (Setd7) in regulating the antioxidant Nrf2 pathway in prostate cancer (PCa) cells and examines the effects of two phytochemicals, phenethyl isothiocyanate (PEITC) and ursolic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service