Skip to Content
Merck
All Photos(1)

Key Documents

244686

Sigma-Aldrich

Sodium

25-35 wt % dispersion in paraffin

Synonym(s):

Sodium metal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Na
CAS Number:
Molecular Weight:
22.99
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

1 mmHg ( 440 °C)

form

solid

reaction suitability

reagent type: reductant

concentration

25-35 wt % dispersion in paraffin

resistivity

4.69 μΩ-cm, 20°C

bp

883 °C (lit.)

mp

97.8 °C (lit.)

SMILES string

[Na]

InChI

1S/Na

InChI key

KEAYESYHFKHZAL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yin Zhang et al.
Experimental and therapeutic medicine, 12(5), 2928-2936 (2016-11-25)
The use of self-expanding metallic stents (SEMS) is the current treatment of choice for malignant gastrointestinal obstructions. A paclitaxel-eluting metallic SEMS (PEMS) may have an antitumor effect on esophageal tissue. PEMS with 10% paclitaxel or conventional SEMS were inserted into
Supriya S Jalimarada et al.
Investigative ophthalmology & visual science, 54(6), 4330-4340 (2013-06-08)
Mutations in SLC4A11, a member of the SLC4 superfamily of bicarbonate transporters, give rise to corneal endothelial cell dystrophies. SLC4A11 is a putative Na⁺ borate and Na⁺:OH⁻ transporter. Therefore we ask whether SLC4A11 in corneal endothelium transports borate (B[OH]₄⁻), bicarbonate
Nicole Li et al.
American heart journal, 166(5), 815-822 (2013-11-02)
Cardiovascular diseases are the leading cause of death and disability in China. High blood pressure caused by excess intake of dietary sodium is widespread and an effective sodium reduction program has potential to improve cardiovascular health. This study is a
Julijana Vuchkova et al.
American journal of physiology. Renal physiology, 307(5), F525-F532 (2014-07-06)
Albuminuria in nephrotic states is thought to arise from the formation of large pores in the glomerular capillary wall as large hydrodynamic probes, like Ficoll, have increased fractional clearance. In the present study, we tested for large pore formation in
Etana Padan
Biochimica et biophysica acta, 1837(7), 1047-1062 (2013-12-24)
The crystal structure of down-regulated NhaA crystallized at acidic pH4 [21] has provided the first structural insights into the antiport mechanism and pH regulation of a Na(+)/H(+) antiporter [22]. On the basis of the NhaA crystal structure [21] and experimental

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service