Skip to Content
Merck
All Photos(1)

Key Documents

660167

Sigma-Aldrich

Sodium silica gel Stage I

extent of labeling: ~35 wt. % Na-K loading

Synonym(s):

Na-SG-I

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
11101527
NACRES:
NA.22

form

solid

Quality Level

autoignition temp.

248-257 °F

reaction suitability

reagent type: reductant

extent of labeling

~35 wt. % Na-K loading

Application

Alternative to other reagents for desulfurizations, dehalogenations(as Wurtz coupling reactions), and Birch reductions.

Additionally can be used as:
  • A reducing agent for aliphatic ester substrates
  • A deprotectant in place of the typical heavy metal deprotectant

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - Water-react 2

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P. F. Vogt and B. Bodnar,
Spec. Chem., 29, 22-24 (2009)
Dye, J. L. et al
Journal of the American Chemical Society, 127, 9339-9339 (2005)
P. Nandi, et al.,
Tetrahedron Letters, 50, 3864-3866 (2009)

Articles

SiGNa Chemistry has recently developed a technology for encapsulating alkali metals into nano-structured porous oxides to create stable, free flowing powders with varying reactivities (based on the intended application).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service