Skip to Content
Merck
All Photos(2)

Key Documents

244864

Sigma-Aldrich

Potassium

chunks (in mineral oil), 98% trace metals basis

Synonym(s):

Atomic potassium, Potassium hyperkalemia

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
K
CAS Number:
Molecular Weight:
39.10
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.09 mmHg ( 260 °C)

Quality Level

Assay

98% trace metals basis

form

chunks (in mineral oil)

reaction suitability

reagent type: reductant

resistivity

6.1 μΩ-cm, 20°C

bp

760 °C (lit.)

mp

64 °C (lit.)

density

0.86 g/mL at 25 °C (lit.)

SMILES string

[K]

InChI

1S/K

InChI key

ZLMJMSJWJFRBEC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Combines under pressure with silver metal to form 2 new compounds: K2Ag and K3Ag.†
Potassium can be used in the synthesis of potassium containing solid-state chalcogenides with promising thermoelectric properties. It may also be used in the dehalogenation of 1,3-di-tert-butyl-2,2-dichloro-1,3-diaza-2-sila-4-cyclopentene to form 1,3-di-tert-butyl-2,3-dihydro-1H-1,3,2-diazasilol-2-ylidene.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Incorporation of A2Q into HgQ and dimensional reduction to A2Hg3Q4 and A2Hg6Q7 (A= K, Rb, Cs; Q= S, Se). Access of Li ions in A2Hg6Q7 through topotactic ion-exchange.
Axtell EA, et al.
Journal of the American Chemical Society, 120(1), 124-136 (1998)
Synthesis and reactivity of a stable silylene.
Haaf M, et al.
Journal of the American Chemical Society, 120(49), 12714-12719 (1998)
Lattice thermal conductivity of K 2 (Bi 1? z Sb z) 8 Se 13 solid solutions.
Kyratsi T, et al.
Journal of Applied Physics, 95(8), 4140-4146 (2004)
Glucose-insulin-potassium revived: current status in acute coronary syndromes and the energy-depleted heart.
Alexandra N Grossman et al.
Circulation, 127(9), 1040-1048 (2013-03-06)
Mayumi Tada et al.
The European journal of neuroscience, 39(11), 1720-1728 (2014-01-11)
Calcium imaging of individual neurons is widely used for monitoring their activity in vitro and in vivo. Synthetic fluorescent calcium indicator dyes are commonly used, but the resulting calcium signals sometimes suffer from a low signal-to-noise ratio (SNR). Therefore, it

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service