Skip to Content
Merck
All Photos(2)

Key Documents

240338

Sigma-Aldrich

2-Pyrrolidinone

≥99%

Synonym(s):

2-Pyrrolidone, Butyrolactam

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7NO
CAS Number:
Molecular Weight:
85.10
Beilstein:
105241
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

2.9 (vs air)

Assay

≥99%

form

solid

refractive index

n20/D 1.487 (lit.)

bp

245 °C (lit.)

mp

23-25 °C (lit.)

solubility

benzene: miscible(lit.)
carbon disulfide: miscible(lit.)
chloroform: miscible(lit.)
diethyl ether: miscible(lit.)
ethanol: miscible(lit.)
ethyl acetate: miscible(lit.)
water: miscible(lit.)

density

1.12 g/mL at 25 °C (lit.)

SMILES string

O=C1CCCN1

InChI

1S/C4H7NO/c6-4-2-1-3-5-4/h1-3H2,(H,5,6)

InChI key

HNJBEVLQSNELDL-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

The FT-IR spectra of the hydrogen bonding lactam 2-pyrrolidinone in CCl4 was studied[1].

Application

2-Pyrrolidinone is used as a starting material in the synthesis of polyvinylpyrrolidone (PVP),[2] (±)-tashiromine,[3] (±)-tetraponerine T4,[4] (+)-ricciocarpins A and B.[5]

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intramolecular Additions of Various ?-Nucleophiles to Chemoselectively Activated Amides and Application to the Synthesis of (?)-Tashiromine.
Belanger G, et al.
The Journal of Organic Chemistry, 71(2), 704-712 (2006)
Application of enantioselective radical reactions: synthesis of (+)-ricciocarpins A and B.
Sibi MP and He L
Organic Letters, 6(11), 1749-1752 (2004)
Photocatalyzed degradation of polymers in aqueous semiconductor suspensions: V. Photomineralization of lactam ring-pendant polyvinylpyrrolidone at titania/water interfaces.
Horikoshi S, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 138(1), 69-77 (2001)
Electrophilic Activation of Lactams with Tf2O and Pyridine: Expedient Synthesis of (?)-Tetraponerine T4.
Charette A B, et al.
Organic Letters, 7(24), 5401-5404 (2005)
Kiran Sankar Maiti et al.
Physical chemistry chemical physics : PCCP, 14(47), 16294-16300 (2012-11-08)
The mid-IR spectrum of the hydrogen bonding lactam 2-pyrrolidinone in CCl(4) was studied using FT-IR spectroscopy accompanied by a quantum chemical anharmonic normal mode analysis combined with a Monte Carlo approach based on semi-empirical harmonic frequencies. We characterized and assigned

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service