V209
δ-Valerolactam
98%
Synonym(s):
delta-Valerolactam, 2-Piperidone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5H9NO
CAS Number:
Molecular Weight:
99.13
Beilstein:
106434
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
bp
256 °C (lit.)
81-82 °C/0.1 mmHg (lit.)
mp
38-40 °C (lit.)
SMILES string
O=C1CCCCN1
InChI
1S/C5H9NO/c7-5-3-1-2-4-6-5/h1-4H2,(H,6,7)
InChI key
XUWHAWMETYGRKB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Jingwei Zhang et al.
ACS synthetic biology, 6(5), 884-890 (2017-04-18)
ε-Caprolactam and δ-valerolactam are important commodity chemicals used in the manufacture of nylons, with millions of tons produced annually. Biological production of these highly valued chemicals has been limited due to a lack of enzymes that cyclize ω-amino fatty acid
T Senda et al.
The Journal of organic chemistry, 66(21), 6852-6856 (2001-10-13)
Catalytic asymmetric synthesis of 4-aryl-2-piperidinones was realized for the first time by asymmetric 1,4-addition of arylboron reagents to 5,6-dihydro-2(1H)-pyridinones in the presence of a chiral bisphosphine-rhodium catalyst. In the reaction introducing 4-fluorophenyl group, the use of 4-fluorophenylboroxine and 1 equiv
P A Reddy et al.
Journal of medicinal chemistry, 40(1), 44-49 (1997-01-03)
A series of 3-substituted 2-piperidinone (delta-valerolactam) and hexahydro-2H-azepin-2-one (epsilon-caprolactam) derivatives were prepared and evaluated as anticonvulsants in mice. In the 2-piperidinone series, 3,3-diethyl compound 7b is the most effective anticonvulsant against pentylenetetrazole-induced seizures (ED50, 37 mg/kg; PI (TD50/ED50), 4.46), and
Moitrayee Mukherjee et al.
The journal of physical chemistry. A, 116(40), 9888-9896 (2012-09-19)
A comparative analysis for relative stability between normal and tautomeric forms in the excited electronic states of 7-azaindole···δ-valerolactam 1:1 complex and 7-azaindole homodimer has been presented. The tautomeric configuration of the complex is estimated to be ~6 kcal/mol more stable
Asymmetric synthesis of gamma-keto-delta-lactam derivatives: application to the synthesis of a conformationally constrained surrogate of Ala-Ser dipeptide.
S D Koulocheri et al.
The Journal of organic chemistry, 66(23), 7915-7918 (2001-11-10)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service