Skip to Content
Merck
All Photos(1)

Key Documents

V3409

Sigma-Aldrich

1-Vinyl-2-pyrrolidinone

contains sodium hydroxide as inhibitor, ≥99%

Synonym(s):

1-Vinyl-2-pyrrolidone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9NO
CAS Number:
Molecular Weight:
111.14
Beilstein:
110513
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

3.8 (vs air)

Quality Level

vapor pressure

0.1 mmHg ( 24 °C)

Assay

≥99%

autoignition temp.

685 °F

contains

sodium hydroxide as inhibitor
100 ppm sodium hydroxide as inhibitor (added to bulk material)

expl. lim.

10 %

refractive index

n20/D 1.512 (lit.)

bp

92-95 °C/11 mmHg (lit.)

density

1.04 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C=CN1CCCC1=O

InChI

1S/C6H9NO/c1-2-7-5-3-4-6(7)8/h2H,1,3-5H2

InChI key

WHNWPMSKXPGLAX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Vinyl-2-pyrrolidinone is a colorless to yellow liquid, with a characteristic odor. Its melting point is around 13.5oC and boils at about 90-92oC. VP is completely miscible in water and in most organic solvents but partially miscible in aliphatic hydrocarbons. Industrial production of VP by reacting 2-pyrrolidone with acetylene at high pressure and temperature has been reported. The vinylation process proceeds in liquid phase and is catalyzed by 3-pyrrolidone -potassium hydroxide.

Application

1-Vinyl-2-pyrrolidinone (VP) is used in the study of macrophotoinitiator characteristics of poly (vinylchloride) type molecules. Also, used to study the genotoxicity of polyvinylpyrrolidone coated silver nanoparticles.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - STOT RE 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kris N J Stevens et al.
Biomaterials, 32(5), 1264-1269 (2010-11-26)
Central venous catheters (CVCs) have become indispensable in the treatment of neonates and patients undergoing chemotherapy or hemodialysis. A CVC provides easy access to the patient's circulation, thus enabling facile monitoring of hemodynamic parameters, nutritional support, or administration of (cytostatic)
Andrew M Telford et al.
Langmuir : the ACS journal of surfaces and colloids, 27(23), 14207-14217 (2011-10-21)
We investigated the dewetting of metastable poly(N-vinylpyrrolidone) (PNVP) thin films (45 nm) on top of polystyrene (PS) thin films (58 nm) as a function of annealing temperature and molecular weight of PS (96 and 6850 kg/mol). We focused on the
Ana C Borges et al.
Acta biomaterialia, 7(9), 3412-3421 (2011-06-10)
The swelling and compressive mechanical behavior as well as the morphology and biocompatibility of composite hydrogels based on Tween® 20 trimethacrylate (T3), N-vinyl-2-pyrrolidone (NVP) and nanofibrillated cellulose (NFC) were assessed in the present study. The chemical structure of T3 was
Synthesis and characterization of poly (vinylchloride) type macrophotoinitiator comprising side-chain thioxanthone via click chemistry.
Akat H and Ozkan M
Express Polymer Letters, 5(4), 318-326 (2011)
Volker Engels et al.
Dalton transactions (Cambridge, England : 2003), 39(28), 6496-6502 (2010-06-23)
A modified polyol-based reduction method in ethylene glycol that incorporates poly(N-vinylpyrrolidone) (PVP, M(av) = 10,000; 40,000; 55,000) as polymeric anti-agglomerant alongside a reducing additive (N(2)H(4) x H(2)O, NaBH(4), NaH(2)PO(2) x H(2)O) has been employed to investigate the influence of synthetic

Articles

Monomers for ophthalmic use aim for purity, reliability, and comfort, driving innovation for affordable contact lenses.

Monomers for ophthalmic use aim for purity, reliability, and comfort, driving innovation for affordable contact lenses.

Monomers for ophthalmic use aim for purity, reliability, and comfort, driving innovation for affordable contact lenses.

Monomers for ophthalmic use aim for purity, reliability, and comfort, driving innovation for affordable contact lenses.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service