Skip to Content
Merck
All Photos(2)

Documents

232076

Sigma-Aldrich

Molybdenum(II) acetate dimer

98%

Synonym(s):

Di-molybdenum tetraacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mo2(OCOCH3)4
CAS Number:
Molecular Weight:
428.06
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

needles

reaction suitability

core: molybdenum

storage temp.

−20°C

SMILES string

CC(=O)O[Mo]OC(C)=O.CC(=O)O[Mo]OC(C)=O

InChI

1S/4C2H4O2.2Mo/c4*1-2(3)4;;/h4*1H3,(H,3,4);;/q;;;;2*+2/p-4

InChI key

DOOLFANBWPPEGQ-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service