Skip to Content
Merck
All Photos(3)

Key Documents

227749

Sigma-Aldrich

Bis(acetylacetonato)dioxomolybdenum(VI)

Synonym(s):

MoO2(acac)2, Molybdenum diacetylacetonate dioxide, Molybdenum dioxide bis(acetylacetonate), Molybdenum dioxydiacetylacetonate, Molybdenyl acetylacetonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3COCH=C(O-)CH3]2MoO2
CAS Number:
Molecular Weight:
326.15
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

powder

reaction suitability

core: molybdenum
reagent type: catalyst

mp

184 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

CC(=O)\C=C(\C)O[Mo](=O)(=O)O\C(C)=C/C(C)=O

InChI

1S/2C5H8O2.Mo.2O/c2*1-4(6)3-5(2)7;;;/h2*3,6H,1-2H3;;;/q;;+2;;/p-2/b2*4-3-;;;

InChI key

SKMUJBBRXZPAJY-VGKOASNMSA-L

General description

Bis(acetylacetonato)dioxomolybdenum(VI) is used as a starting material and catalyst for oxidative transformation reactions.

Application

Bis(acetylacetonato)dioxomolybdenum(VI) can be used:
  • As a starting material to synthesize dioxomolybdenum(VI) Schiff base complexes, applicable as catalysts in the oxidation of alcohols using H2O2.
  • To prepare graphene oxide supported heterogeneous molybdenum catalysts for epoxidation of olefins.
  • As a catalyst in the etherification and thio-etherification reactions of thiols and alcohols.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Priecel et al.
Frontiers in chemistry, 8, 216-216 (2020-04-24)
This investigation deals with NiMo-alumina hydrotreating catalysts effective in the deoxygenation of rapeseed oil. The main goal was to compare catalyst structure and their deoxygenation performance and to link these parameters to reveal important structural information regarding the catalyst's intended
Covalent functionalization of graphene oxide with molybdenum-carboxylate complexes: new reusable catalysts for the epoxidation of olefins
Masteri-Farahani M and Mirshekar S
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 538(1), 387-392 (2018)
Dinuclear and polynuclear dioxomolybdenum (VI) Schiff base complexes: synthesis, structural elucidation, spectroscopic characterization, electrochemistry and catalytic property
Ngan NK, et al.
Polyhedron, 33(1), 235-251 (2012)
Synthesis, structures, electrochemical studies and antioxidant activities of cis-dioxomolybdenum (VI) complexes of the new bisthiocarbohydrazones
Kaya Y, et al.
Journal of Molecular Structure, 1102, 17-126 (2015)
Molybdenum (VI)-catalyzed dehydrative construction of CO and CS bonds formation via etherification and thioetherification of alcohols and thiols
Singh RR, et al.
Molecular Catalysis, 492(1), 110954-110954 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service