Skip to Content
Merck
All Photos(1)

Key Documents

220523

Sigma-Aldrich

(2-Methylbutyl)amine

≥97%

Synonym(s):

(±)-1-Amino-2-methylbutane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(CH3)CH2NH2
CAS Number:
Molecular Weight:
87.16
Beilstein:
1718806
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97%

refractive index

n20/D 1.4116 (lit.)

bp

94-97 °C (lit.)

solubility

alcohols and water: soluble(lit.)

density

0.738 g/mL at 25 °C (lit.)

SMILES string

CCC(C)CN

InChI

1S/C5H13N/c1-3-5(2)4-6/h5H,3-4,6H2,1-2H3

InChI key

VJROPLWGFCORRM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

46.4 °F - closed cup

Flash Point(C)

8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Bai et al.
Journal of food science, 84(3), 481-489 (2019-02-19)
Alcohols, aldehydes, ketones, amines, and sulfur compounds are essential aroma compounds related to fish flavor and spoilage. Gas chromatography-mass spectrometry (GC-MS) is an instrument that is widely used to identify and quantify volatile and semi-volatile compounds in fish products. In
James M Brimson et al.
Neurotoxicity research, 34(2), 263-272 (2018-03-29)
Alzheimer's disease is a neurodegenerative disease that affects 44 million people worldwide, costing the world $605 billion to care for those affected not taking into account the physical and psychological costs for those who care for Alzheimer's patients. Dipentylammonium is
Jianping Zhao et al.
Phytochemistry, 152, 191-203 (2018-05-22)
Sceletium tortuosum (Aizoaceae) is widely recognised for the treatment of stress, anxiety and depression, as well as for obsessive compulsive disorders. A comprehensive intraspecies chemotypic variation study, using samples harvested from two distinct regions of South Africa, was done using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service