Skip to Content
Merck
All Photos(1)

Key Documents

463779

Sigma-Aldrich

Ruthenium(III) chloride hydrate

99.98% trace metals basis

Synonym(s):

Trichlororuthenium hydrate, Ruthenium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
RuCl3 · xH2O
CAS Number:
Molecular Weight:
207.43 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.98% trace metals basis
form:
powder and chunks

Quality Level

Assay

99.98% trace metals basis

form

powder and chunks

reaction suitability

reagent type: catalyst
core: ruthenium

impurities

≤250.0 ppm Trace Metal Analysis

SMILES string

O.Cl[Ru](Cl)Cl

InChI

1S/3ClH.H2O.Ru/h3*1H;1H2;/q;;;;+3/p-3

InChI key

BIXNGBXQRRXPLM-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Ruthenium(III) chloride hydrate exhibits excellent catalytic activity, attributed to the redox properties of ruthenium suitable for fuel cells and other electrochemical devices. It has ability to facilitate charge transfer and exhibit photoactive behavior.

Application

Ruthenium(III) chloride hydrate can be used:
  • As a precursor to prepare ultralow Ru loaded graphene aerogel that exhibits electrocatalytic performance in the hydrogen evolution reaction (HER), oxygen evolution reaction (OER), and oxygen reduction reaction (ORR).
  • As a dopant to prepare Ru-doped TiO2 electron transport layer for mesoscopic perovskite solar cell by spin-coating technique. This helps to enhance photoconversion efficiency and air stability.
  • To fabricate ultra-uniform Ru nanocluster catalyst via small-molecule self-assembly pyrolysis method. These catalysts exhibit excellent hydrogen evolution activity in Zinc air batteries under all pH conditions.

Other Notes

soluble form

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fujita, S.-I., et al.
J. Catal., 255, 95-95 (2004)
Cho, C.S. et al.
Tetrahedron Letters, 40, 1499-1499 (1999)
Won-Hee Kim et al.
Organic letters, 8(12), 2543-2545 (2006-06-02)
An efficient oxidant-free oxidation for a wide range of alcohols was achieved by a recyclable ruthenium catalyst. The catalyst was prepared from readily available reagents by a one-pot synthesis through nanoparticle generation and gelation. [structure: see text]
Yao Zhang et al.
European journal of medicinal chemistry, 86, 449-455 (2014-09-10)
Ruthenium-based anticancer complexes have become increasingly popular for study over the last two decades. Although ruthenium complexes are currently being investigated in clinical trials, there are still some difficulties with their delivery and associated side effects. Human serum albumin (HSA)-based
Huaiyi Huang et al.
Dalton transactions (Cambridge, England : 2003), 44(35), 15602-15610 (2015-08-08)
Ruthenium complexes have been considered as promising substitutes for cisplatin in cancer chemotherapy. However, novel ruthenium-based therapies are faced with some limitations, such as unimpressive cytotoxicity toward solid tumors. Herein, we designed and synthesized phenyl-substituted terpyridyl ruthenium(ii) complexes ([Ru(tpy)(bpy)Cl](+) (Ru1)

Articles

Hydrogen is one of the most important resources in providing food, fuel, and chemical products for our everyday life. Sustainable catalytic hydrogen production from bioethanol has gained significant attention in recent years due to globally diminishing fossil fuel supplies, which have necessitated the search for new chemical feedstocks.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service