Skip to Content
Merck
All Photos(2)

Documents

208523

Sigma-Aldrich

Ruthenium(III) chloride

Ru content 45-55%

Synonym(s):

Ruthenium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
RuCl3
CAS Number:
Molecular Weight:
207.43
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

core: ruthenium
reagent type: catalyst
reaction type: Atom Transfer Radical Polymerization (ATRP)

density

3.11 g/mL at 25 °C (lit.)

SMILES string

Cl[Ru](Cl)Cl

InChI

1S/3ClH.Ru/h3*1H;/q;;;+3/p-3

InChI key

YBCAZPLXEGKKFM-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Ruthenium(III) chloride is a chemical compound, that can be used as a mild Lewis acid catalyst for the acetalization of aldehydes, acetalization of alcohols, and conversion of ketoximes to amides. Additionally, it can also be used as a precursor to synthesize Ru nanoparticles.

Application

Ruthenium(III) chloride is used as a catalyst:

  • In the synthesis of β‐amino alcohols by nucleophilic opening of epoxides with anilines.
  • In the acetylation of varies of phenols, alcohols, thiols, and amines under mild conditions.
  • In the synthesis of α‐aminonitriles by mixing aldehydes, amines, and trimethylsilyl cyanides.

Other Notes

insoluble form

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shū Kobayashi et al.
Organic letters, 4(8), 1319-1322 (2002-04-13)
Several transition metal salts were found to catalyze aza-Michael reactions of enones with carbamates efficiently. The catalytic activity was strongly dependent on the nature of the metal salts. While conventional Lewis acids such as BF(3).OEt(2), AlCl(3), or TiCl(4) showed lower
Ruthenium (III) chloride catalyzed acylation of alcohols, phenols, thiols, and amines
De SK
Tetrahedron Letters, 45(14), 2919-2922 (2004)
Sébastien Perdriau et al.
ChemSusChem, 5(12), 2427-2434 (2012-10-13)
Cardanol, a constituent of cashew nutshell liquid (CNSL), was subjected to transfer hydrogenation catalyzed by RuCl(3) using isopropanol as a reductant. The side chain of cardanol, which is a mixture of a triene, a diene, and a monoene, was selectively
Ruthenium (III) chloride-catalyzed ring opening of epoxides with aromatic amines
De SK and Gibbs RA
Synthetic Communications, 35(20), 2675-2680 (2005)
Takaya Terashima et al.
Macromolecular rapid communications, 33(9), 833-841 (2012-04-25)
Multifunctional microgel-core star polymers with ruthenium catalysts are designed as catalyst-bearing nanoreactors to improve activity, controllability, and functionality tolerance in living radical polymerization. Multifunctional ligands are efficiently incorporated into the core of star polymers by sequential tandem procedures: 1) ruthenium-catalyzed

Articles

Micro review of reversible addition/fragmentation chain transfer (RAFT) polymerization.

Micro review of reversible addition/fragmentation chain transfer (RAFT) polymerization.

Micro review of reversible addition/fragmentation chain transfer (RAFT) polymerization.

Micro review of reversible addition/fragmentation chain transfer (RAFT) polymerization.

Protocols

We presents an article featuring procedures that describe polymerization of methyl methacrylate and vinyl acetate homopolymers and a block copolymer as performed by researchers at CSIRO.

We present an article about RAFT, or Reversible Addition/Fragmentation Chain Transfer, which is a form of living radical polymerization.

Polymerization via ATRP procedures demonstrated by Prof. Dave Haddleton's research group at the University of Warwick.

Polymerization via ATRP procedures demonstrated by Prof. Dave Haddleton's research group at the University of Warwick.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service