Skip to Content
Merck
All Photos(1)

Key Documents

220329

Sigma-Aldrich

7-Methylcoumarin

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8O2
CAS Number:
Molecular Weight:
160.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

solid

bp

171-172 °C/11 mmHg (lit.)

mp

128-130 °C (lit.)

functional group

ester

SMILES string

Cc1ccc2C=CC(=O)Oc2c1

InChI

1S/C10H8O2/c1-7-2-3-8-4-5-10(11)12-9(8)6-7/h2-6H,1H3

InChI key

DLHXRDUXNVEIEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Antimitotic potential of 7-methylcoumarin has been investigated in Allium sativum promeristem1.

Application

7-Methylcoumarin was used as internal standard during the simultaneous determination of furocoumarin compounds in rat plasma by HPLC.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Risto O Juvonen et al.
Xenobiotica; the fate of foreign compounds in biological systems, 46(1), 14-24 (2015-06-13)
1. Information about the metabolism of compounds is essential in drug discovery and development, risk assessment of chemicals and further development of predictive methods. 2. In vitro and in silico methods were applied to evaluate the metabolic and inhibitory properties of 6-methylcoumarin
K Ishihara et al.
Journal of chromatography. B, Biomedical sciences and applications, 753(2), 309-314 (2001-05-04)
A simple and sensitive column-switching HPLC method was developed for the simultaneous determination of two furocoumarin compounds, byak-angelicin and oxypeucedanin hydrate, which are the main components of hot water extract of Angelica dahurica root (AE), in rat plasma. Plasma sample

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service