Skip to Content
Merck
All Photos(3)

Key Documents

H24003

Sigma-Aldrich

Umbelliferone

99%

Synonym(s):

7-Hydroxy-2H-1-benzopyran-2-one, 7-Hydroxycoumarin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6O3
CAS Number:
Molecular Weight:
162.14
Beilstein:
127683
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

mp

230 °C (dec.) (lit.)

solubility

dioxane: soluble
ethanol: soluble

SMILES string

Oc1ccc2C=CC(=O)Oc2c1

InChI

1S/C9H6O3/c10-7-3-1-6-2-4-9(11)12-8(6)5-7/h1-5,10H

InChI key

ORHBXUUXSCNDEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Umbelliferone can be used:
  • To synthesize fluorescent probes to detect cyanide anions in water at biological pH and for in vitro and in vivo detection of hydrogen peroxide.
  • In the preparation of coumarin-based polymer as optical data storage material.
  • As a precursor to synthesize novobiocin derivatives that exhibit antibacterial agents as well as inhibitors of DNA gyrase.
  • As the starting material to synthesize (+)-decursinol and (+)-angelmarin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorescent chemodosimeter for selective detection of cyanide in water.
Lee K S, et al.
Organic Letters, 10(1), 49-51 (2008)
Reversible two-photon optical data storage in coumarin-based copolymers.
Iliopoulos K, et al.
Journal of the American Chemical Society, 132(41), 14343-14345 (2010)
Novobiocin: redesigning a DNA gyrase inhibitor for selective inhibition of hsp90.
Burlison J A, et al.
Journal of the American Chemical Society, 128(48), 15529-15536 (2006)
Total synthesis of (+)-angelmarin.
Magolan J and Coster M J
The Journal of Organic Chemistry, 74(14), 5083-5086 (2009)
Rational design of a fluorescent hydrogen peroxide probe based on the umbelliferone fluorophore.
Du L, et al.
Tetrahedron Letters, 49(19), 3045-3048 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service