Skip to Content
Merck
All Photos(1)

Documents

W515809

Sigma-Aldrich

7-Methoxycoumarin

≥98%

Synonym(s):

Herniarin, Methyl umbelliferyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8O3
CAS Number:
Molecular Weight:
176.17
Beilstein:
141728
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic

Quality Level

Assay

≥98%

mp

115-121 °C
117-121 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

balsam

SMILES string

COc1ccc2C=CC(=O)Oc2c1

InChI

1S/C10H8O3/c1-12-8-4-2-7-3-5-10(11)13-9(7)6-8/h2-6H,1H3

InChI key

LIIALPBMIOVAHH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

7-Methoxycoumarin is one of the main volatile odorant found in key lime essential oil and tarragon leaves.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Impact of estragole and other odorants on the flavour of anise and tarragon
Zeller A and Rychlik M
Flavour and Fragrance Journal, 22(2), 105-113 (2007)
Characterization of aroma volatiles in key lime essential oils (Citrus aurantifolia Swingle).
Chisholm MG, et al.
Flavour and Fragrance Journal, 18(2), 106-115 (2003)
W Legrum et al.
The Journal of pharmacology and experimental therapeutics, 221(3), 790-794 (1982-06-01)
Pretreatment of rats with cobaltous chloride has been shown previously to reduce the content of cytochrome P-450 in the hepatic microsomal protein. This is accompanied by a corresponding decrease in substrate oxidation, e.g. ethyl morphine demethylation, in vitro. The present
Stefan Starcević et al.
Journal of medicinal chemistry, 54(1), 248-261 (2010-12-09)
17β-Hydroxysteroid dehydrogenase type 1 (17β-HSD1) is an enzyme that catalyzes NADPH-dependent reduction of the weak estrogen, estrone, into the most potent estrogen, estradiol, which exerts proliferative effects via the estrogen receptors. Overexpression of 17β-HSD1 in estrogen-responsive tissues is related to
Prince Firdoos Iqbal et al.
European journal of medicinal chemistry, 44(5), 2252-2259 (2008-08-05)
In continuation of our search for potential antiamoebic agents from folklore Indian medicinal plants, we found that the benzene and ethyl acetate extracts from the root bark of Adina cordifolia exhibited strong antiamoebic activity with IC(50) values of 2.92 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service