Skip to Content
Merck
All Photos(4)

Documents

189464

Sigma-Aldrich

Poly(ethylene oxide)

average MV 4,000,000 (nominal), powder, hydroxyl, BHT as inhibitor

Synonym(s):

Polyethylene oxide, PEO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(-CH2CH2O-)n
CAS Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.23

product name

Poly(ethylene oxide), average Mv ~4,000,000 (nominal), contains <1000 ppm BHT as inhibitor

form

powder

Quality Level

mol wt

average Mv ~4,000,000 (nominal)

contains

<1000 ppm BHT as inhibitor
100-1000 ppm BHT as inhibitor

viscosity

1,650-5,500 cP, 1 % in H2O(25 °C, Brookfield)(lit.)

transition temp

Tm 65 °C

density

1.21 g/mL at 25 °C

functional group

hydroxyl

SMILES string

[H]OCCO

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly (ethylene oxide) (PEO) is a crystalline thermoplastic polymer. It is usually available over a wide range of molecular weights; the higher molecular weight entities are called poly (ethylene oxide), poly-oxyethylene or polyoxirane.

Application

PEO may be used to afford PEO-salt complexes which are act as polymeric electrolytes to be used for alkali metal rechargeable batteries. PEO-polyester micelles have been reportedly used for drug delivery. PEO hydrogels have been reported to be used for culture media, replacing agar. Cross-linked PEO may be used for drug delivery and blood filtration. Compounds form association complexes with high molecular weight PEO; for example:

  • bromine combines with PEO in controlled release,
  • carboxymethyl dextran and carechol tannis form complexes with PEO to act as adhesives,
  • copolymer of methyl vinyl ether and maleic anhydride form complex with PEO for its use in microencapsulation of non aqueous inks.

Industrially, PEO may be used to modify bituminous emulsion in paving compositions and as a dispersant for calamine lotion.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hart DS
Polyethylene Oxide-polyester Micelles for Drug Delivery null
Whelan T
Polymer Technology Dictionary null
FE Jr Bailey
Poly(Ethylene Oxide) null
Bailey FE
Alkylene Oxides and Their Polymers null
Yi Huang et al.
Polymers, 12(5) (2020-05-21)
Controlling the rheological property of suspensions consisting of colloidal particles and polymers is necessary in industry. Especially, gels induced by shear (shake-gel) are interesting phenomena in rheological field. To gain insight into the shake-gel phenomena of the aqueous suspensions of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service