Skip to Content
Merck
All Photos(1)

Key Documents

188344

Sigma-Aldrich

3-(Trifluoromethyl)benzoic acid

99%

Synonym(s):

α,α,α-Trifluoro-m-toluic acid, 3-Carboxybenzotrifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4CO2H
CAS Number:
Molecular Weight:
190.12
Beilstein:
2049239
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

238.5 °C/775 mmHg (lit.)

mp

104-106 °C (lit.)

SMILES string

OC(=O)c1cccc(c1)C(F)(F)F

InChI

1S/C8H5F3O2/c9-8(10,11)6-3-1-2-5(4-6)7(12)13/h1-4H,(H,12,13)

InChI key

FQXQBFUUVCDIRK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

pKa values of 3-(trifluoromethyl)benzoic acid in water and in methanol has been measured. Solubility of 3-(trifluoromethyl)benzoic acid in dense carbon dioxide was evaluated to investigate the influence of fluorination on the solubility of organic pharmaceuticals in dense carbon dioxide.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aneela Maalik et al.
Bioorganic chemistry, 88, 102946-102946 (2019-05-06)
An irrefutable advancement has been noted for the infectious diseases caused due to ureolytic bacteria through the development of various drugs. Keeping in mind the extremely valuable synthetic utility and medicinal significance of thiourea derivatives, synthesis of new 3-trifluoromethyl benzoic
Solubility of fluorinated pharmaceuticals in dense carbon dioxide.
Laitinen A, et al.
Organic Process Research & Development, 4(5), 353-356 (2000)
K H Engesser et al.
Archives of microbiology, 149(3), 188-197 (1988-01-01)
The TOL plasmid-encoded enzymes of the methylbenzoate pathway in Pseudomonas putida mt-2 cometabolized 3-trifluoromethyl (TFM)-benzoate. Two products, 3-TFM-1,2-dihydroxy-2-hydrobenzoate (3-TFM-DHB) and 2-hydroxy-6-oxo-7,7,7-trifluoro-hepta-2,4-dienoate (7-TFHOD) were identified chemically and by spectroscopic properties. TFM-substituted analogues of the metabolites of the methylbenzoate pathway were generally
S A Selifonov et al.
Biochemical and biophysical research communications, 213(3), 759-767 (1995-08-24)
Pseudomonas aeruginosa strain 142 oxidizes 2-halobenzoates via a multicomponent oxygenase (V. Romanov and R.P. Hausinger, J. Bacteriol., 1994, 176(11), 3368-3374). The intermediacy of a highly unstable cis-diol in the reaction has been proposed. Direct evidence for this is currently lacking
B F Taylor et al.
FEMS microbiology letters, 110(2), 213-216 (1993-06-15)
m- and p-trifluoromethyl (TFM)-benzoates are incompletely degraded by aerobic bacteria that catabolize alkylbenzoates; biodegradation ceases after ring-fission with the accumulation of a trifluoromethyl muconate semialdehyde (2-hydroxy-6-oxo-7,7,7-trifluorohepta-2,4-dienoate, TFHOD) which is resistant to biochemical attack. A bacterium (Strain V-1), isolated from sea-water

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service