187879
2,5-Dimethoxybenzyl alcohol
99%
Synonym(s):
(2,5-Dimethoxyphenyl)methanol, 1-(Hydroxymethyl)-2,5-dimethoxybenzene, 2,5-Dimethoxybenzenemethanol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3O)2C6H3CH2OH
CAS Number:
Molecular Weight:
168.19
Beilstein:
1947607
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
refractive index
n20/D 1.547 (lit.)
bp
122-125 °C/1 mmHg (lit.)
density
1.173 g/mL at 25 °C (lit.)
functional group
hydroxyl
SMILES string
COc1ccc(OC)c(CO)c1
InChI
1S/C9H12O3/c1-11-8-3-4-9(12-2)7(5-8)6-10/h3-5,10H,6H2,1-2H3
InChI key
WGQMUABRZGUAOS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
2,5-Dimethoxybenzyl alcohol was used as starting reagent in the synthesis of [(S)S]-3,6-dimethoxy-2-(p-tolylsulfinyl)-benzaldehyde. It was used in the synthesis of daunornycinone. It was also used in the preparation of pillar[n]arenes (n = 5 or 6) via cyclooligomerization reaction with an appropriate Lewis acid catalyst.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Diastereodivergent additions of aluminum and magnesium reagents to [(S)S]-3, 6-dimethoxy-2-(p-tolylsulfinyl)-benzaldehyde.
Almori'n A, et al.
Tetrahedron Letters, 44(30), 5597-5600 (2003)
An exceptional 5-endo-trig reversal; a convergent synthesis of daunomycinone.
Keay BA and Rodrigo R.
Canadian Journal of Chemistry, 61(3), 637-639 (1983)
Preparation of Pillar [n] arenes by Cyclooligomerization of 2, 5-Dialkoxybenzyl Alcohols or 2, 5-Dialkoxybenzyl Bromides.
Ma Y, et al.
European Journal of Organic Chemistry, 2011(27), 5331-5335 (2011)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service