Skip to Content
Merck
All Photos(1)

Key Documents

150355

Sigma-Aldrich

4-tert-Butylbenzoic acid

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H4CO2H
CAS Number:
Molecular Weight:
178.23
Beilstein:
607545
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

mp

162-165 °C (lit.)

functional group

carboxylic acid

SMILES string

CC(C)(C)c1ccc(cc1)C(O)=O

InChI

1S/C11H14O2/c1-11(2,3)9-6-4-8(5-7-9)10(12)13/h4-7H,1-3H3,(H,12,13)

InChI key

KDVYCTOWXSLNNI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-tert-Butylbenzoic acid was determined in water samples by means of liquid chromatography-electrospray ionisation mass spectrometry (LC-ESI-MS).

Application

4-tert-Butylbenzoic acid was used as a potent yeast sirtuin (Sir2p) inhibitor.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Repr. 1B - STOT RE 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi-Pei Chen et al.
Bioorganic & medicinal chemistry letters, 24(1), 349-352 (2013-11-26)
Employing a genetically modified yeast strain as a screening tool, 4-dimethylaminobenzoic acid (5) was isolated from the marine sediment-derived Streptomyces sp. CP27-53 as a weak yeast sirtuin (Sir2p) inhibitor. Using this compound as a scaffold, a series of disubstituted benzene
Patrycja Makoś et al.
Journal of chromatography. A, 1517, 26-34 (2017-09-01)
The paper presents a new method for the determination of 15 carboxylic acids in samples of postoxidative effluents from the production of petroleum bitumens using ion-pair dispersive liquid-liquid microextraction and gas chromatography coupled to mass spectrometry with injection port derivatization.
Inhibition of hepatic gluconeogenesis and lipogenesis by benzoic acid, p-tert.-butylbenzoic acid, and a structurally related hypolipidemic agent SC-33459.
S A McCune et al.
Archives of biochemistry and biophysics, 214(1), 124-133 (1982-03-01)
I Anundi et al.
Chemico-biological interactions, 50(3), 277-288 (1984-08-01)
Isolated hepatocytes incubated with selenite (30-100 microM) exhibited changes in the glutathione redox system as shown by an increase in O2 consumption, oxidation of glutathione and loss of NADPH. Selenite (50 microM) raised O2 consumption within the 1 h and
Adeline Martin et al.
Carbohydrate polymers, 93(2), 718-730 (2013-03-19)
The aim of this work was to develop the formation of multilayered coating incorporating a cyclodextrin polyelectrolyte onto a non-woven polyethylene terephthalate (PET) textile support in order to obtain reservoir and sustained release properties towards bioactive molecules. We optimized the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service