Skip to Content
Merck
All Photos(4)

Key Documents

H53704

Sigma-Aldrich

N-Hydroxyphthalimide

97%, for peptide synthesis

Synonym(s):

2-Hydroxy-1H-isoindole-1,3(2H)-dione, 2-Hydroxyisoindole-1,3-dione, 2-Hydroxyphthalimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H5NO3
CAS Number:
Molecular Weight:
163.13
Beilstein:
131208
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Product Name

N-Hydroxyphthalimide, 97%

Quality Level

Assay

97%

form

crystalline

reaction suitability

reaction type: Addition Reactions

mp

233 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

ON1C(=O)c2ccccc2C1=O

InChI

1S/C8H5NO3/c10-7-5-3-1-2-4-6(5)8(11)9(7)12/h1-4,12H

InChI key

CFMZSMGAMPBRBE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For preparation of amine nucleosides for coupling into non-anionic, anti-sense oligonucleosides.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 33, 2645-2645 (1992)
Francesca D'Acunzo et al.
European journal of biochemistry, 269(21), 5330-5335 (2002-10-24)
To investigate how solubility and steric issues affect the laccase-catalysed oxidation of phenols, a series of oligomeric polyphenol compounds, having increasing size and decreasing solubility in water, was incubated with laccase. The extent of substrate conversion, and the nature of
Application of a dual linker with a reference cleavage site to discover a new reaction between amines and N-hydroxyphthalimide.
Viktor Krchnák et al.
Journal of combinatorial chemistry, 7(4), 523-525 (2005-07-12)
Riyuan Lin et al.
Organic letters, 14(16), 4158-4161 (2012-08-03)
A metal-free N-hydroxyphthalimide (NHPI) catalyzed aerobic oxidative cleavage of olefins has been developed. Molecular oxygen is used as the oxidant and reagent for this oxygenation reaction. This methodology has prevented the use of toxic metals or overstoichiometric amounts of traditional
Yuuki Amaoka et al.
The Journal of organic chemistry, 77(22), 9959-9969 (2012-11-02)
A direct conversion of C(sp(3))-H bonds to C(sp(3))-N bonds has been achieved by utilizing catalytic N-hydroxyphthalimide (NHPI) and stoichiometric dialkyl azodicarboxylate. NHPI functions as a precursor of the electron-deficient phthalimide N-oxyl radical (PINO) to abstract hydrogens, and dialkyl azodicarboxylate acts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service