H19808
3-Hydroxybenzaldehyde
≥99%
Synonym(s):
3-Formylphenol, 5-Hydroxybenzaldehyde, m-Formylphenol, m-Hydroxybenzaldehyde
Sign Into View Organizational & Contract Pricing
All Photos(5)
About This Item
Linear Formula:
HOC6H4CHO
CAS Number:
Molecular Weight:
122.12
Beilstein:
507099
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥99%
bp
191 °C/50 mmHg (lit.)
mp
100-103 °C (lit.)
SMILES string
Oc1cccc(C=O)c1
InChI
1S/C7H6O2/c8-5-6-2-1-3-7(9)4-6/h1-5,9H
InChI key
IAVREABSGIHHMO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
3-Hydroxybenzaldehyde can be used as a reactant along with ethyl acetoacetate and thiourea in the synthesis of corresponding dihydropyrimidine-2-thione (monastrol), using Yb(OTf)3 as a catalyst by Biginelli cyclocondensation reaction.
It can also be used as a starting material for the synthesis of:
It can also be used as a starting material for the synthesis of:
- (-)-Quinocarcin, isoxazolo[3,4-e][2,1]benzisoxazole and 3-n-propylphenol.
- Oligo-3-hydroxybenzaldehyde (O-3HBA) by oxidative polycondensation.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
A practical synthesis of 3-n-propylphenol, a component of tsetse fly attractant blends
Ujvary I and Mikite G
Organic Process Research & Development, 7(4), 585-587 (2003)
Improved synthesis and preparative scale resolution of racemic monastrol
Dondoni A, et al.
Tetrahedron Letters, 43(34), 5913-5916 (2002)
A novel cyclization to isoxazolo [3, 4-e][2, 1] benzisoxazole
Katritzky AR, et al.
Tetrahedron Letters, 43(18), 3449-3451 (2002)
T Suthan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 87, 194-198 (2011-12-24)
Novel organic nonlinear optical (NLO) material 3-hydroxybenzaldehyde (3HBA) single crystals have been grown by modified vertical Bridgman technique using the single and double wall ampoules with nano translation. Single crystal X-ray diffraction analysis reveals that 3HBA crystallizes in orthorhombic system
The synthesis and thermal properties of oligo-3-hydroxybenzaldehyde synthesized by oxidative polycondensation
Mart H, et al.
Journal of Applied Polymer Science, 101(2), 892-897 (2006)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service