Skip to Content
Merck
All Photos(1)

Key Documents

159549

Sigma-Aldrich

4-(Bromomethyl)benzoic acid

97%

Synonym(s):

α-Bromo-p-toluic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2C6H4CO2H
CAS Number:
Molecular Weight:
215.04
Beilstein:
1862870
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

224-229 °C (lit.)

functional group

bromo
carboxylic acid

SMILES string

OC(=O)c1ccc(CBr)cc1

InChI

1S/C8H7BrO2/c9-5-6-1-3-7(4-2-6)8(10)11/h1-4H,5H2,(H,10,11)

InChI key

CQQSQBRPAJSTFB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

4-(Bromomethyl)benzoic acid was used in the chemical modification of 5,10,15,20-tetra(m-hydroxyphenyl)chlorin (temoporfin), second generation photosensitizer. It was also used in the synthesis of 4-(5-arylidene-2,4-dioxothiazolidin-3-yl) methylbenzoic acids.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chendong Ji et al.
Chemistry, an Asian journal, 11(16), 2316-2321 (2016-07-14)
Electrospun ultrathin fiber-based sensors are desirable because of their practicality and sensitivity. Ammonia-detection systems are in high demand in different areas, including the industrial and agricultural fields. However, current technologies rely on large and complex instruments that restrict their actual
Marcela S Lopes et al.
European journal of medicinal chemistry, 46(11), 5443-5447 (2011-09-24)
A series of nitroaromatic compounds was synthesized and evaluated as potential antileishmanial and trypanocidal agents. Five compounds exerted significant anti-leishmanial activity in vitro against promastigotes forms of Leishmania (L.) amazonensis, with IC(50) in the range of 23-59 μmol L(-1), but
Rosanna Maccari et al.
Bioorganic & medicinal chemistry, 15(15), 5137-5149 (2007-06-05)
4-(5-Arylidene-2,4-dioxothiazolidin-3-yl)methylbenzoic acids (2) were synthesized and evaluated in vitro as inhibitors of PTP1B and LMW-PTP, two protein tyrosine phosphatases (PTPs) which act as negative regulators of the metabolic and mitotic signalling of insulin. The synthesis of compounds 2 represents an
Q Yu et al.
Chemical communications (Cambridge, England), 50(81), 12150-12153 (2014-09-02)
LiYF4:Tm(3+)/Yb(3+) upconverting nanoparticles (UCNPs) were functionalized with the second generation photosensitizer 5,10,15,20-tetra(m-hydroxyphenyl)chlorin (m-THPC, Temoporfin, Foscan®). m-THPC was modified using 4-(bromomethyl)benzoic acid, which induced a bathochromic shift of the m-THPC blue absorption peak. The nanoconstruct causes up to 70% cell death
Salvatore Lombardo et al.
Langmuir : the ACS journal of surfaces and colloids, 33(22), 5473-5481 (2017-05-13)
The interaction of bovine serum albumin (BSA) with sulfated, carboxylated, and pyridinium-grafted cellulose nanocrystals (CNCs) was studied as a function of the degree of substitution by determining the adsorption isotherm and by directly measuring the thermodynamics of interaction. The adsorption

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service