Skip to Content
Merck
All Photos(2)

Key Documents

105899

Sigma-Aldrich

Cyclohexanol

ReagentPlus®, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11OH
CAS Number:
Molecular Weight:
100.16
Beilstein:
906744
EC Number:
MDL number:
UNSPSC Code:
12352104
eCl@ss:
39023102
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
bp:
160-161 °C (lit.)
vapor pressure:
0.98 mmHg ( 25 °C)

grade

reagent

Quality Level

vapor density

3.5 (vs air)

vapor pressure

0.98 mmHg ( 25 °C)

product line

ReagentPlus®

Assay

99%

autoignition temp.

572 °F

expl. lim.

12.25 %

dilution

(for general lab use)

refractive index

n20/D 1.465 (lit.)

bp

160-161 °C (lit.)

mp

20-22 °C (lit.)

density

0.948 g/mL at 25 °C (lit.)

SMILES string

OC1CCCCC1

InChI

1S/C6H12O/c7-6-4-2-1-3-5-6/h6-7H,1-5H2

InChI key

HPXRVTGHNJAIIH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyclohexanol is an alicyclic alcohol. It is industrially produced from phenol, via hydrogenation in the presence of Ni/Al2O3 catalyst. It is also produced from cyclohexane, via oxidation with molecular oxygen in the presence of Co3O4 nanocrystals (catalyst). A mixture of cyclohexanone (K) and cyclohexanol (A) is known as KA oil. KA oil can be generated from cyclohexane, via liquid-phase oxidation.

Application

Cyclohexanol may be used in the preparation of ε-caprolactam and adipic acid, important precursors for the industrial synthesis of nylon-6 and nylon-6.6, respectively.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

147.2 °F - closed cup - DIN 51794

Flash Point(C)

64 °C - closed cup - DIN 51794

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nazanin Vafaei et al.
Applied biochemistry and biotechnology, 191(3), 905-920 (2020-01-11)
The time course study of high monoester mixtures from soybean oil (HMMS) synthesis, as healthier alternatives to trans food products, in a supercritical CO2 (SCCO2) medium with and without enzyme, was investigated. Phosphorous nuclear magnetic resonance (31P-NMR) was used to
Cong Gao et al.
International journal of biological macromolecules, 162, 1642-1652 (2020-08-17)
The low cost, environmental friendliness, and reproducibility of kraft lignin (KL) make it a potential candidate for the development of new green material. The phosphorylation of KL can extend its application as a flame-retardant material. Herein, the phosphorylated kraft lignin
Catalytic oxidation of cyclohexane to cyclohexanol and cyclohexanone over Co3O4 nanocrystals with molecular oxygen.
Zhou L, et al.
Applied Catalysis A: General, 292, 223-228 (2005)
Eagleson M.
Concise Encyclopedia Chemistry, 511-511 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 811-811 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service