Skip to Content
Merck
All Photos(1)

Key Documents

8.01641

Sigma-Aldrich

Benzoyl peroxide

(with 25% H2O) for synthesis

Synonym(s):

Benzoyl peroxide, Dibenzoyl peroxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10O4
CAS Number:
Molecular Weight:
242.23
MDL number:
UNSPSC Code:
12352120
EC Index Number:
202-327-6
NACRES:
NA.22

vapor pressure

<1 hPa ( 20 °C)

Quality Level

form

crystals

autoignition temp.

>380 °C

potency

7710 mg/kg LD50, oral (Rat)

reaction suitability

reagent type: oxidant

mp

100-105 °C (decomposition)

density

1.33 g/cm3 at 25 °C

bulk density

500‑600 kg/m3

functional group

peroxide

storage temp.

15-25°C

SMILES string

O(OC(=O)c2ccccc2)C(=O)c1ccccc1

InChI

1S/C14H10O4/c15-13(11-7-3-1-4-8-11)17-18-14(16)12-9-5-2-6-10-12/h1-10H

InChI key

OMPJBNCRMGITSC-UHFFFAOYSA-N

General description

Benzoyl peroxide is an organic peroxide commonly used to initiate free radical polymerizations and to form copolymers through grafting reactions. It is also used as an oxidizer during chemical reactions.

Application

Benzoyl peroxide is used:

  • In the direct N–O bond synthesis from 1,2-diamines.
  • As an initiator in the synthesis of the cross-linked unsaturated copolymer by cross-linking methyl methacrylate with Poly (2-butene maleate) polyester.

Analysis Note

Assay (iodometric): 72,0 - 77,0 %(m)
Melting range (after drying): 102 - 105 °C
Identity (IR): conforms

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Org. Perox. C - Skin Sens. 1A

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of new substituted bis (benzoyl) peroxides under aqueous conditions in the presence of stearyltrimethylammonium chloride
Moorhoff CM, et al.
Monatshefte fur Chemie / Chemical Monthly, 135(4), 397-405 (2004)
Direct N--O bond formation via oxidation of amines with benzoyl peroxide
Banerjee A, et al.
Chemical Science, 10(71), 2124-2129 (2019)
Preparation A Cross-Linked Copolymer By Using Unsaturated Poly (2-Butene Maleate) Polyester and Methyl Methacrylate
Saltan F
Celal Bayar University Journal of Science, 18(3), 279-285 (2022)
Effect of benzoyl peroxide on the ultrasonic degradation of poly (vinyl acetate
Madras G, et al.
Polymer Degradation and Stability, 73(1), 33-38 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service