Skip to Content
Merck
All Photos(1)

Key Documents

156388

Sigma-Aldrich

Pentafluorophenylhydrazine

97%

Synonym(s):

(Perfluorophenyl)hydrazine, NSC 88334, PFPH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6F5NHNH2
CAS Number:
Molecular Weight:
198.09
Beilstein:
747800
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

mp

74-76 °C (lit.)

functional group

fluoro
hydrazine

SMILES string

NNc1c(F)c(F)c(F)c(F)c1F

InChI

1S/C6H3F5N2/c7-1-2(8)4(10)6(13-12)5(11)3(1)9/h13H,12H2

InChI key

BYCUWCJUPSUFBX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

The product has been used to determine the purity of N-tert-butyl-N-[1-diethylphosphono-(2,2-dimethylpropyl)]nitroxide (DEPN) using 1H NMR spectroscopy.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elena E Stashenko et al.
Journal of chromatographic science, 44(6), 347-353 (2006-08-04)
Specific chromatographic methodologies are developed for the analysis of carboxylic acids (C(2)-C(6), benzoic) and aldehydes (C(2)-C(10)) of low molecular weight in diverse matrices, such as air, automotive exhaust gases, human breath, and aqueous matrices. For carboxylic acids, the method is
Michael Otteneder et al.
Chemical research in toxicology, 15(3), 312-318 (2002-03-19)
Malondialdehyde is a ubiquitous product of lipid peroxidation that reacts with DNA to form premutagenic lesions. Principal among them is pyrimido-[1,2-alpha]purin-10(3H)-one (M(1)G). M(1)G has recently been found to be a reactive electrophile in DNA that couples with amines at basic
S Rochat et al.
Journal of chromatography. A, 1216(36), 6424-6432 (2009-08-05)
The GC-SNIF technique was used to obtain the olfactograms of shrimps, and their impact odorants were identified by MS hyphenated to the GC/olfactometric system and by comprehensive two-dimensional GC hyphenated to a time-of-flight MS. Confirming these identifications by their linear
A sensitive determination of alpha-keto acids by gas-liquid chromatography and its application to the assay of L-glutamate dehydrogenase and aminotransferases.
S Ohmori et al.
Analytical biochemistry, 112(2), 204-212 (1981-04-01)
Xiaobing Pang et al.
Talanta, 85(1), 406-414 (2011-06-08)
The classical analytical method for gaseous carbonyl measurements based on solid sorbent coated with 2,4-dinitrophenylhydrazine (DNPH) and analysis by HPLC/UV suffers from limited resolution of carbonyls with similar molecular structures and high molecular weights. In this paper, we report the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service