Skip to Content
Merck
All Photos(1)

Key Documents

683973

Sigma-Aldrich

(5aR,10bS)-5a,10b-Dihydro-2-(2,4,6-trimethylphenyl)-4H,6H-indeno[2,1-b]-1,2,4-triazolo[4,3-d]-1,4-oxazinium chloride monohydrate

93%

Synonym(s):

Bode Catalyst 2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22ClN3O · H2O
CAS Number:
Molecular Weight:
385.89
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

93%

form

solid

optical activity

[α]20/D +158°, c = 1 in chloroform

mp

226-230 °C

functional group

ether

SMILES string

[Cl-].Cc1cc(C)c(c(C)c1)-[n+]2c[n@@H]3[C@@H]4[C@@H](Cc5ccccc45)OCc3n2

InChI

1S/C21H22N3O.ClH/c1-13-8-14(2)20(15(3)9-13)24-12-23-19(22-24)11-25-18-10-16-6-4-5-7-17(16)21(18)23;/h4-9,12,18,21H,10-11H2,1-3H3;1H/q+1;/p-1/t18-,21+;/m1./s1

InChI key

GUECWMLEUCWYOS-WKOQGQMTSA-M

General description

It is an N-mesityl-substituted chiral triazolium N-heterocyclic carbene (NHC) compound. It was developed as an organocatalyst by the Bode group, for the catalytic generation of reactive species including activated carboxylates, homoenolates, and enolates.

Application

(5aR,10bS)-5a,10b-Dihydro-2-(2,4,6-trimethylphenyl)-4H,6H-indeno[2,1-b]-1,2,4-triazolo[4,3-d]-1,4-oxazinium chloride monohydrate can be used as a catalyst:
  • In the preparation of dihydropyridinones by reacting enals or α′-hydroxyenones with vinylogous amides via aza-Claisen annulations.
  • In the oxidative coupling reactions of di(hetero)arylmethanes with enals to yield benzimidazole fused lactams.
  • In the synthesis of organosilanes by reacting enals with β-silyl enones.

Legal Information

Sold in collaboration with BioBlocks, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N-Heterocyclic Carbene (NHC) Compounds
Aldrich Chemfiles, 7(9), 21-21 (2007)
Carbene-catalyzed enantioselective oxidative coupling of enals and di (hetero) arylmethanes
Chen Q, et al.
Chemical Science, 9(46), 8711-8715 (2018)
Enantioselective synthesis of dihydropyridinones via NHC-catalyzed aza-Claisen reaction
Wanner B, et al.
Organic Letters, 13(19), 5378-5381 (2011)
Access to Enantioenriched Organosilanes from Enals and β-Silyl Enones: Carbene Organocatalysis
Zhang Y, et al.
Angewandte Chemie (International Edition in English), 130(17), 4684-4688 (2018)
Synthesis of a N-Mesityl Substituted Aminoindanol-Derived Triazolium Salt.
Struble JR and Bode JW.
Organic Syntheses, 362-376 (2010)

Articles

Rovis has demonstrated that triazolium salt in the presence of a base can act as an N-heterocyclic carbene organocatalyst1 in highly enantioselective intramolecular Stetter reactions.

Related Content

Bode Group creates N-mesityl-NHC catalysts for enantioselective annulations and SnAP reagents for one-pot aldehyde conversion into N-heterocycles.

Bode Group creates N-mesityl-NHC catalysts for enantioselective annulations and SnAP reagents for one-pot aldehyde conversion into N-heterocycles.

Bode Group creates N-mesityl-NHC catalysts for enantioselective annulations and SnAP reagents for one-pot aldehyde conversion into N-heterocycles.

Bode Group creates N-mesityl-NHC catalysts for enantioselective annulations and SnAP reagents for one-pot aldehyde conversion into N-heterocycles.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service