Skip to Content
Merck
All Photos(3)

Documents

C4026

Sigma-Aldrich

Sodium hexanoate

99-100%

Synonym(s):

Caproic acid sodium salt, Hexanoic acid sodium salt, Sodium caproate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4COONa
CAS Number:
Molecular Weight:
138.14
Beilstein:
3631527
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

99-100%

form

powder

lipid type

saturated FAs

storage temp.

2-8°C

SMILES string

[Na+].CCCCCC([O-])=O

InChI

1S/C6H12O2.Na/c1-2-3-4-5-6(7)8;/h2-5H2,1H3,(H,7,8);/q;+1/p-1

InChI key

UDWXLZLRRVQONG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lee-Mei Ng et al.
Journal of bioscience and bioengineering, 122(5), 550-557 (2016-10-26)
Aquitalea sp. USM4 (JCM 19919) was isolated from a freshwater sample at Lata Iskandar Waterfall in Perak, Malaysia. It is a rod-shaped, gram-negative bacterium with high sequence identity (99%) to Aquitalea magnusonii based on 16S rRNA gene analysis. Aquitalea sp.
Huating Li et al.
Diabetes, 61(4), 797-806 (2012-02-18)
Fibroblast growth factor 21 (FGF21) stimulates fatty acid oxidation and ketone body production in animals. In this study, we investigated the role of FGF21 in the metabolic activity of sodium butyrate, a dietary histone deacetylase (HDAC) inhibitor. FGF21 expression was
Su Li et al.
Biophysical journal, 101(3), 700-708 (2011-08-03)
Low-molecular-weight carboxylic acids have many properties common to small molecule drugs. The transport of these acids across cell membranes has been widely studied, but these studies have produced wildly varying permeability values. Recent reports have even claimed that the transport
Noelia Luque et al.
Journal of chromatography. A, 1235, 84-91 (2012-03-17)
In this work, we propose a microextraction method based on a new supramolecular solvent (SUPRAS) made up of reverse aggregates of hexanoic acid, combined with liquid chromatography/triple quadrupole mass spectrometry (LC/QQQ MS-MS) for the determination of the perfluoroalkyl substances (PFASs)
Zhana Kravchuk et al.
Journal of plant physiology, 168(4), 359-366 (2010-10-19)
Soil drench treatments with hexanoic acid can effectively protect Arabidopsis plants against Botrytis cinerea through a mechanism based on a stronger and faster accumulation of JA-dependent defenses. Plants impaired in ethylene, salicylic acid, abscisic acid or glutathion pathways showed intact

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service