Skip to Content
Merck
All Photos(2)

Documents

115959

Sigma-Aldrich

Benzoyl cyanide

98%

Synonym(s):

Phenylglyoxylonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCN
CAS Number:
Molecular Weight:
131.13
Beilstein:
1072101
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

206 °C (lit.)

mp

28-31 °C (lit.)

density

1.106 g/mL at 25 °C (lit.)

SMILES string

O=C(C#N)c1ccccc1

InChI

1S/C8H5NO/c9-6-8(10)7-4-2-1-3-5-7/h1-5H

InChI key

GJQBHOAJJGIPRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzoyl cyanide is used to study the mechanism of reduction of benzoyl cyanide in acetonitrile, N,N-dimethylformamide and acetonitrile. It undergoes hydrolysis by Rhodococcus sp. CCZU10-1 to form benzoylformic acid.

Application

Reagent for selective acylation of amino compounds.

related product

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Konstantin N Bulygin et al.
RNA (New York, N.Y.), 22(2), 278-289 (2015-12-15)
Translation termination in eukaryotes is mediated by release factors: eRF1, which is responsible for stop codon recognition and peptidyl-tRNA hydrolysis, and GTPase eRF3, which stimulates peptide release. Here, we have utilized ribose-specific probes to investigate accessibility of rRNA backbone in
Yi Xu et al.
Journal of virology, 92(11) (2018-03-09)
Translational readthrough of the stop codon of the capsid protein (CP) open reading frame (ORF) is used by members of the Luteoviridae to produce their minor capsid protein as a readthrough protein (RTP). The elements regulating RTP expression are not
Weisi Xiong et al.
Biotechnology letters, 37(8), 1703-1709 (2015-04-22)
To develop a biphasic system for use in plant tissue-mediated biotransformations to overcome the low water solubilities of substrates and inhibitory effects of products. Commonly-used organic solvents and ionic liquid were assayed using the biphasic system to reduce water-insoluble benzoyl
Norma A Macías-Ruvalcaba et al.
The Journal of organic chemistry, 72(2), 589-594 (2007-01-16)
The mechanism of reduction of benzoyl cyanide, 6, p-methoxybenzoyl cyanide, 7, and p-chlorobenzoyl cyanide, 8, has been studied in acetonitrile (6 and 7), N,N-dimethylformamide (6), and acetonitrile containing water (all three compounds). The reaction proceeds by initial reduction to form
Boris Zinshteyn et al.
Nucleic acids research, 47(1), 43-55 (2018-11-27)
Chemical probing methods are crucial to our understanding of the structure and function of RNA molecules. The majority of chemical methods used to probe RNA structure report on Watson-Crick pairing, but tertiary structure parameters such as solvent accessibility can provide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service