Merck
  • An easy-to-use excimer fluorescence derivatization reagent, 2-chloro-4-methoxy-6-(4-(pyren-4-yl)butoxy)-1,3,5-triazine, for use in the highly sensitive and selective liquid chromatography analysis of histamine in Japanese soy sauces.
An easy-to-use excimer fluorescence derivatization reagent, 2-chloro-4-methoxy-6-(4-(pyren-4-yl)butoxy)-1,3,5-triazine, for use in the highly sensitive and selective liquid chromatography analysis of histamine in Japanese soy sauces.
Analytica chimica acta (2015-06-21)
Tatsuki Nakano, Kenichiro Todoroki, Yasuhiro Ishii, Chiemi Miyauchi, Arpaporn Palee, Jun Zhe Min, Koichi Inoue, Kuniaki Suzuki, Toshimasa Toyo'oka
ABSTRACT
In this study, a novel pre-column excimer fluorescence derivatization reagent, 2-chloro-4-methoxy-6-(4-(pyren-4-yl)butoxy)-1,3,5-triazine (CMPT), was developed for polyamines, specifically histamine. By CMPT derivatization, the polyamines, histamine and tyramine were converted to polypyrene derivatives, and emitted intra-molecular excimer fluorescence at 475nm. This could clearly be distinguished from the normal fluorescence emitted from reagent blanks at 375 nm. Unlike conventional excimer fluorescence derivatization reagents, CMPT is chemically stable and its reactivity sustained over at least 36 days even in solution state. We successfully applied this reagent to the sensitive and selective analysis of histamine in different kinds of Japanese commercial soy sauces. The detection and quantification limits of histamine were 15 and 50 μg L(-1), respectively, equating to 1.35 pmol and 4.5 pmol for a 6 μL injection. This sensitivity helped the direct analysis of soy sauce samples only treated by one-step liquid-liquid extraction without concentration. The histamine levels of commercial soy sauce samples (koikuchi, usukuchi and saishikomi) investigated were 1.24-768.5 mg L(-1).
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
2,4,6-Trimethylpyridine, ReagentPlus®, 99%
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
1,8-Diaminooctane, 98%
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Phenethylamine, 99%
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
2,4,6-Trimethylpyridine, puriss. p.a., 99% (GC)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
N-Benzylmethylamine, 97%
Sigma-Aldrich
1,4-Diaminobutane, 99%
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Cadaverine, 95%
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Phenethylamine, purified by redistillation, ≥99.5%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Phenethylamine, ≥99%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis