Skip to Content
Merck
All Photos(4)

Key Documents

225789

Sigma-Aldrich

Chloroform-d

Chloroform-d
1 of 1 reviewers received a sample product or took part in a promotion

99.8 atom % D, contains 0.03 % (v/v) TMS

Synonym(s):

Deuterochloroform

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CDCl3
CAS Number:
Molecular Weight:
120.38
Beilstein:
1697633
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

isotopic purity

99.8 atom % D

Quality Level

Assay

≥99% (CP)

form

liquid

contains

0.03 % (v/v) TMS

technique(s)

NMR: suitable

impurities

≤0.0100% water
water

refractive index

n20/D 1.444 (lit.)

bp

60.9 °C (lit.)

mp

−64 °C (lit.)

density

1.500 g/mL at 25 °C (lit.)

mass shift

M+1

SMILES string

[2H]C(Cl)(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H/i1D

InChI key

HEDRZPFGACZZDS-MICDWDOJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chloroform-d is a deuterated NMR solvent useful in NMR-based research and analyses.

Application

Chloroform-d may be used in the synthesis of dichlorofluoromethane-d (DCFM).[1]

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dichlorofluoromethane-d: a versatile solvent for VT-NMR experiments.
Siegel JS & Anet, FA
The Journal of Organic Chemistry, 53(11), 2629-2630 (1988)
Salvatore Barreca et al.
Natural product research, 33(7), 947-955 (2015-11-28)
This preliminary study has mainly focused on the wax identification by nuclear magnetic resonance (NMR) and removal. Wax is used for many purposes in the field of art as protective coatings on wooden, stone or metal objects. From the comparison
Isotopic dilution studies of the chloroform-chloroform-d system by Raman difference spectroscopy.
Laane J and Kiefer W.
J. Chem. Phys. , 73(10), 4971-4975 (1980)
Quantitative Study of the Bonding of Chloroform-d in Various Solvents by Infrared Spectrometry1.
Lord RC, et al.
Journal of the American Chemical Society, 77(5), 1365-1368 (1955)
Infrared Intensity of the C-D Stretch of Chloroform-d in Various Solvents.
Huggins CM and Pimentel GC.
J. Chem. Phys. , 23(5), 896-898 (1955)

Related Content

Widest range of NMR solvents with excellent chemical purity and the highest isotopic enrichment possible.

NMR spectroscopy elucidates molecular structure and purity via nuclear spin states in a strong magnetic field.

NMR spectroscopy elucidates molecular structure and purity via nuclear spin states in a strong magnetic field.

NMR spectroscopy elucidates molecular structure and purity via nuclear spin states in a strong magnetic field.

See All

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. Cambridge
    • Reviews 7
    • Votes 15
    5 out of 5 stars.

    CDCl3

    Product quality is good. Trace amount of warer is present

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service