Skip to Content
Merck
All Photos(1)

Key Documents

431915

Sigma-Aldrich

Chloroform-d

"100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer

Synonym(s):

Deuterochloroform

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CDCl3
CAS Number:
Molecular Weight:
120.38
Beilstein:
1697633
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

99.96 atom % D

Assay

≥99.0%

form

liquid

contains

0.5 wt. % silver wire as stabilizer

IVD

for in vitro diagnostic use

technique(s)

NMR: suitable

impurities

≤0.0100% water
water

refractive index

n20/D 1.444 (lit.)

bp

60.9 °C (lit.)

mp

−64 °C (lit.)

density

1.500 g/mL at 25 °C (lit.)

mass shift

M+1

SMILES string

[2H]C(Cl)(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H/i1D

InChI key

HEDRZPFGACZZDS-MICDWDOJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chloroform-d (deuterochloroform, CDCl3) is a deuterated NMR solvent useful in NMR-based research and analyses. It may be synthesized by reacting trichloroacetophenone and sodium deuteroxide. The influence of the number of pulses on its multiphoton decomposition (MPD) has been studied. Infrared spectral studies of C-D stretching and bending motions of CDCl3 in different solvents have been investigated. The effect of chloroform-d on the interaction of water with activated carbons has been analyzed. The dependence of molecular reorientation of deuterated chloroform on pressure and temperature has been studied by Raman spectroscopy.

Application

Chloroform-d may be used in the synthesis of dichlorofluoromethane-d.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raman study of molecular reorientation in liquid chloroform and chloroform-d under high pressure.
Schroeder J, et al.
J. Chem. Phys. , 69(12), 5479-5488 (1978)
Dichlorofluoromethane-d: a versatile solvent for VT-NMR experiments.
Siegel JS and Anet FAL.
The Journal of Organic Chemistry, 53(11), 2629-2630 (1988)
Isotope Effect on Dissociation Probabilities in the Mass Spectra of Chloroform and Chloroform-d.
Dibeler VH and Bernstein RB.
J. Chem. Phys. , 19(4), 404-406 (1951)
The effect of collisions in the multiphoton decomposition of chloroform-d.
McRae GA, et al.
J. Chem. Phys. , 92(10), 5997-6003 (1990)
Influence of organics on the structure of water adsorbed on activated carbons.
Turov VV, et al.
Journal of Colloid and Interface Science, 253(1), 23-34 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service